Skip to Content
Merck
All Photos(2)

Key Documents

SAB4503289

Sigma-Aldrich

Anti-GPR101 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

GPR101, Probable G-protein coupled receptor 101

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 56 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GPR101(83550)

General description

Anti-GPR101 Antibody detects endogenous levels of total GPR101 protein.
G protein-coupled receptor 101 (GPR101) codes for an an orphan G protein-coupled receptor. It is expressed at high level in the hypothalamus. The GPR101 gene is mapped to human chromosome Xq26.3.

Immunogen

The antiserum was produced against synthesized peptide derived from human GPR101.

Immunogen Range: 451-500

Biochem/physiol Actions

Mutation in G protein-coupled receptor 101 (GPR101) gene result in acromegaly. The GPR101 protein is expected to play an important role in hypothalamic control of energy homeostasis. Duplication of the GPR101 gene leads to X-linked acrogigantism (XLAG).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Very low frequency of germline GPR101 genetic variation and no biallelic defects with AIP in a large cohort of patients with sporadic pituitary adenomas.
Lecoq AL, et al.
European Journal of Endocrinology, 174(4), 523-530 (2016)
Giampaolo Trivellin et al.
Endocrine-related cancer, 23(5), 357-365 (2016-03-11)
Cushing disease (CD) in children is caused by adrenocorticotropic hormone (ACTH)-secreting pituitary adenomas. Germline or somatic mutations in genes such as MEN1, CDKIs, AIP, and USP8 have been identified in pediatric CD, but the genetic defects in a significant percentage
Gigantism: X-linked acrogigantism and GPR101 mutations.
Iacovazzo D and Korbonits M
Growth Hormone & IGF Research, 30-31, 64-69 (2016)
Germline or somatic GPR101 duplication leads to X-linked acrogigantism: a clinico-pathological and genetic study.
Iacovazzo D, et al.
Acta Neuropathologica Communications, 4(1), 56-56 (2016)
Fady Hannah-Shmouni et al.
Journal of clinical medicine, 11(8) (2022-04-24)
Overgrowth due to growth hormone (GH) excess affects approximately 10% of patients with neurofibromatosis type 1 (NF1) and optic pathway glioma (OPG). Our aim is to describe the clinical, biochemical, pathological, and genetic features of GH excess in a retrospective

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service