Skip to Content
Merck
All Photos(3)

Key Documents

SAB4503194

Sigma-Aldrich

Anti-Calmodulin antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CALM, CALM1, CALM2, CALM3, CAM

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 16 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CALM1(801)

General description

Anti-Calmodulin Antibody detects endogenous levels of total Calmodulin protein.
Calmodulin (CaM), a highly conserved Ca2+-binding protein, is mapped to human chromosome 14q32. Calmodulin is a small protein abundantly present in the brain and coded by three genes (CALM1-3).

Immunogen

The antiserum was produced against synthesized peptide derived from human Calmodulin.

Immunogen Range: 46-95

Application

Anti-Calmodulin antibody has been used in western blotting.

Biochem/physiol Actions

Calmodulin (CaM) possesses Ca2+-dependent modulatory roles. It participates in cell proliferation and muscle activity. Calmodulin plays a major role in catecholaminergic polymorphic ventricular tachycardia (CPVT) and in long QT syndrome (LQTS). This protein modulates the transcriptional processes required for proper heart function.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Orthologs of seven genes (AKT1, CDC42BPB, DIO3, EIF5, JAG2, KLC, NDUFB1) from human chromosome 14q32 map to distal chicken chromosome 5.
Boyd Y, et al.
Mammalian Genome, 13(2), 120-122 (2002)
Presynaptic Calmodulin targets: lessons from structural proteomics.
Lipstein N, et al.
Expert Review of Proteomics, 14(3), 223-242 (2017)
Elucidating arrhythmogenic mechanisms of long-QT syndrome CALM1-F142L mutation in patient-specific induced pluripotent stem cell-derived cardiomyocytes.
Rocchetti M, et al.
Cardiovascular Research, 113(5), 531-541 (2017)
Chunhong Song et al.
BioMed research international, 2017, 8459287-8459287 (2018-01-25)
The aim of this study was to explore the mechanism underlying the antidepression activity of paeoniflorin, the main active ingredient of paeony extract and Shuyu capsules, and determine its effect on the calmodulin/calmodulin-dependent protein kinase II (CaM/CaMKII) signalling pathway and
Ruizhen Li et al.
Molecular biology of the cell, 27(21), 3273-3283 (2016-11-02)
Intracellular calcium transients are a universal phenomenon at fertilization and are required for egg activation, but the exact role of Ca2+ in second-polar-body emission remains unknown. On the other hand, similar calcium transients have not been demonstrated during oocyte maturation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service