Skip to Content
Merck
All Photos(2)

Key Documents

SAB4502583

Sigma-Aldrich

Anti-Retinoblastoma antibody produced in rabbit

affinity isolated antibody

Synonym(s):

P105-RB, PP105, PP110, RB-1, RB1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 106 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RB1(5925)

General description

The gene encoding Retinoblastoma-associated protein (RB1) is a tumor suppressor gene localized on human chromosome 13q14.2. Anti-Retinoblastoma Antibody detects endogenous levels of total Retinoblastoma protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human Retinoblastoma.

Immunogen Range: 771-820

Biochem/physiol Actions

Retinoblastoma-associated protein (RB1) modulates the cell cycle. It acts as a repressor of transcription mediated by E2 factor (E2F) and influences transcriptional silencing. RB1 modulates mitochondrial activities and has a role in the organisation of heterochromatin. Mutations in the gene encoding this protein have been associated with childhood retinoblastoma.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deficiencies in Cellular Processes Modulated by the Retinoblastoma Protein Do Not Account for Reduced Human Cytomegalovirus Replication in Its Absence.
VanDeusen HR and Kalejta RF
Journal of Virology, 89(23), 11965-11974 null
Saeeda Kalsoom et al.
Molecular vision, 21, 1085-1092 (2015-09-24)
Retinoblastoma (RB) is a rare intraocular malignant tumor of the developing retina with an estimated incidence of 1:20,000 live births in children under the age of 5 years. In addition to the abnormal whitish appearance of the pupil or leukocoria
Frequent deletions at 11q23 and 13q14 in B cell prolymphocytic leukemia (B-PLL).
Lens D
Leukemia, 14(3), 427-430 (2000)
Halena R VanDeusen et al.
Journal of virology, 89(9), 5012-5021 (2015-02-20)
The retinoblastoma (Rb) tumor suppressor controls cell cycle, DNA damage, apoptotic, and metabolic pathways. DNA tumor virus oncoproteins reduce Rb function by either inducing Rb degradation or physically disrupting complexes between Rb and its myriad binding proteins. Human cytomegalovirus (HCMV)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service