Skip to Content
Merck
All Photos(2)

Documents

SAB4502329

Sigma-Aldrich

Anti-AMPK α antibody produced in rabbit

affinity isolated antibody

Synonym(s):

5′-AMP-activated protein kinase, AAPK1, AMPK α-1 chain, AMPK-α1, catalytic α-1 chain

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 62 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AAPK1(5562)

General description

Anti-AMPK α antibody detects endogenous levels of total AMPK α protein.
The AMPK α (protein kinase AMP-activated catalytic subunit α1) gene is located on human chromosome 5p13.1. The gene is evolutionarily conserved.

Immunogen

The antiserum was produced against synthesized peptide derived from human AMPK alpha.

Immunogen Range: 140-189

Application

Anti-AMPK α antibody produced in rabbit has been used in immunohistochemistry.

Biochem/physiol Actions

The AMPK α (protein kinase AMP-activated catalytic subunit α1) gene is associated with the regulation of cell cycle, cell proliferation, energy metabolism and inhibition of rapamycin-pathway. Therefore, it plays an important role in carcinogenesis. From recent studies, variation in the gene is known to be involved in gastric cancer. AMPK α might induce anti-inflammatory response in adipose tissue, upon its activation resulting in a beneficial effect in obesity. Decreased cellular ATP or AMP is known to stimulate AMPK enzyme. Activated AMPK stimulates pathways such as mitochondrial biogenesis, glucose transport in muscle and mitochondrial biogenesis, in order to replenish cellular ATP. On the other hand, it prevents anabolic pathways that consume ATP including biosynthesis of fatty acid and cholesterol as well as adipogenesis. The AMPK signaling pathway is generally stimulated in response to stress, energy deficiency and/or oxygen inadequacy.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

AMPK and HIF signaling pathways regulate both longevity and cancer growth: the good news and the bad news about survival mechanisms.
Salminen A
Biogerontology, 17(4), 655-680 (2016)
Additive interactions between PRKAA1 polymorphisms and Helicobacter pylori CagA infection associated with gastric cancer risk in Koreans.
Eom SY
Cancer Medicine, 5(11), 3236-3335 (2016)
A genome-wide association study identifies new susceptibility loci for non-cardia gastric cancer at 3q13.31 and 5p13.1.
Shi Y
Nature Genetics, 43(12), 1215-1218 (2011)
Activation of AMP-activated protein kinase rapidly suppresses multiple pro-inflammatory pathways in adipocytes including IL-1 receptor-associated kinase-4 phosphorylation.
Mancini SJ
Molecular and Cellular Endocrinology, 440, 44-56 (2017)
Interaction of renin-angiotensin system and adenosine monophosphate-activated protein kinase signaling pathway in renal carcinogenesis of uninephrectomized rats.
Yang KK
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine, 39(5) (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service