Skip to Content
Merck
All Photos(2)

Key Documents

SAB4502249

Sigma-Aldrich

Anti-Keratin 20 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CK-20, Cytokeratin-20, K20, Keratin type I cytoskeletal 20

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 48 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KRT20(54474)

General description

Anti-Keratin 20 Antibody detects endogenous levels of total Keratin 20 protein.
The gene KRT20 (keratin 20) is mapped to human chromosome 17. The gene encodes an intermediate filament which is mainly expressed in the gastrointestinal epithelial cells and epidermal Merkel cells. The protein is a type 1 keratin.

Immunogen

The antiserum was produced against synthesized peptide derived from human Keratin 20.

Immunogen Range: 375-424

Biochem/physiol Actions

KRT20 (keratin 20) helps in intermediate filament organization. In metastatic colorectal cancer, it can be used as a biomarker for checking circulating tumor cells.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angélica Morales-Miranda et al.
Molecular and clinical oncology, 8(5), 694-698 (2018-05-05)
Approximately 15% of colorectal cancer (CRC) cases exhibit microsatellite instability (MSI), which appears to be associated with unique biological behavior. The present study presents a case of appendiceal carcinoma associated with MSI that responded well to adjuvant chemotherapy. Clinical, pathological
Sara C Shalin et al.
The American Journal of dermatopathology, 36(11), 904-906 (2014-06-06)
Merkel cell carcinoma (MCC) is recognized by its morphologic features and by its classic immunophenotypic properties. Although MCCs demonstrating nonclassic immunoreactivities have been described, a case documenting a change in immunophenotype during the course of disease progression has not been
M Lacroix et al.
The International journal of biological markers, 17(1), 5-23 (2002-04-09)
Breast cancer remains a major cause of death in women from Western countries. In the near future, advances in both nucleic acids technology and tumor biology should be widely exploited to improve the diagnosis, prognosis, and outcome prediction of this
Charlotte Welinder et al.
Cancer letters, 358(1), 43-46 (2014-12-22)
Cytokeratin 20 (CK20) is a well-established marker for colon epithelium. Herein, we suggest that CK20 is a biomarker for detecting circulating tumor cells (CTCs) in patients with metastatic colorectal cancer. Blood specimens (7.5 mL) were collected during surgery after liver mobilization
Qin Zhou et al.
Molecular biology of the cell, 14(7), 2959-2971 (2003-07-15)
Of the >20 epithelial keratins, keratin 20 (K20) has an unusual distribution and is poorly studied. We began to address K20 function, by expressing human wild-type and Arg80-->His (R80H) genomic (18 kb) and cDNA K20 in cells and mice. Arg80

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service