Skip to Content
Merck
All Photos(3)

Key Documents

SAB4502148

Sigma-Aldrich

Anti-PDGFR β antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CD140b, PDGF-R-β, PDGFR, PDGFR-β, kinase PDGFR-β

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 123 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PDGFRB(5159)

General description

PDGFRB (platelet-derived growth factor receptor β) is a type III tyrosine kinase receptor, which exists as a homodimer. This receptor interacts with multiple ligands, but its main physiological ligand is PDGF-BB homodimer. It is located on human chromosome 5q32.

Immunogen

The antiserum was produced against synthesized peptide derived from human PDGFR beta.

Immunogen Range: 991-1040

Biochem/physiol Actions

PDGFRB (platelet-derived growth factor receptor β), upon interaction with PGDFB, regulates multiple processes such as, sclerotic disorders, wound healing and tumor angiogenesis. Gain-of-function mutation in this gene is linked with Penttinen syndrome. Loss-of-function mutation in this gene results in the neurodegenerative disorder primary familial brain calcification (PFBC).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Massive thromboembolism owing to the left ventricular thrombus associated with the hypereosinophilic syndrome
Kim J S, et al.
The Korean journal of thoracic and cardiovascular surgery, 47(5), 478-478 (2014)
Hongmei Duan et al.
Neural regeneration research, 19(2), 409-415 (2023-07-25)
Attempts have been made to use cell transplantation and biomaterials to promote cell proliferation, differentiation, migration, and survival, as well as angiogenesis, in the context of brain injury. However, whether bioactive materials can repair the damage caused by ischemic stroke
Michael Vanlandewijck et al.
PloS one, 10(11), e0143407-e0143407 (2015-11-26)
Primary Familial Brain Calcification (PFBC), a neurodegenerative disease characterized by progressive pericapillary calcifications, has recently been linked to heterozygous mutations in PDGFB and PDGFRB genes. Here, we functionally analyzed several of these mutations in vitro. All six analyzed PDGFB mutations
Jennifer J Johnston et al.
American journal of human genetics, 97(3), 465-474 (2015-08-19)
Penttinen syndrome is a distinctive disorder characterized by a prematurely aged appearance with lipoatrophy, epidermal and dermal atrophy along with hypertrophic lesions that resemble scars, thin hair, proptosis, underdeveloped cheekbones, and marked acro-osteolysis. All individuals have been simplex cases. Exome

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service