Skip to Content
Merck
All Photos(2)

Key Documents

SAB4501849

Sigma-Aldrich

Anti-MDM2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Double minute 2 protein, HDM2, Oncoprotein Mdm2, Ubiquitin-protein ligase E3 Mdm2, p53-binding protein Mdm2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA
IF
WB
clone:
polyclonal
species reactivity:
human, mouse
citations:
6
technique(s):
ELISA: 1:1000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 55 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MDM2(4193)

General description

Anti-MDM2 antibody detects endogenous levels of total MDM2 protein.
Murine double minute 2 (MDM2) is an oncogenic protein. It is an E3 ubiquitin-protein ligase, that is mapped to human chromosome 12q15.

Immunogen

The antiserum was produced against synthesized peptide derived from human MDM2.

Immunogen Range: 132-181

Application

Anti-MDM2 antibody has been used in immunohistochemistry.

Biochem/physiol Actions

Murine double minute 2 (MDM2) is a negative regulator of the p53 tumor suppressor. It regulates cell cycle, cellular differentiation, cell fate determination, DNA repair and transcription. Epithelial-mesenchymal transition and metastasis of ovarian cancer SKOV3 cells are induced by MDM2.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Introduction of the MDM2 T309G Mutation in Primary Human Retinal Epithelial Cells Enhances Experimental Proliferative Vitreoretinopathy
Zhou G, et al.
Investigative Ophthalmology & Visual Science, 58(12), 5361-5367 (2017)
Angom Ranjana Devi et al.
Indian journal of clinical biochemistry : IJCB, 36(3), 296-303 (2021-07-06)
Nicotine, responsible for the addictive properties of tobacco, is widely used in nicotine replacement therapy for tobacco use cessation. We investigated the time-dependent effect of treatment with nicotine on the tumor suppressor, DNA repair and immune responses. Swiss Albino mice
MDM2 promotes epithelial-mesenchymal transition and metastasis of ovarian cancer SKOV3 cells
Chen Y, et al.
British Journal of Cancer, 117(8), 1192-1192 (2017)
HIV-1 Tat potently stabilises Mdm2 and enhances viral replication
Raja R, et al.
The Biochemical Journal (2017)
HMGA2 is the partner of MDM2 in well-differentiated and dedifferentiated liposarcomas whereas CDK4 belongs to a distinct inconsistent amplicon
Italiano A, et al.
International Journal of Cancer. Journal International Du Cancer, 122(10), 2233-2241 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service