Skip to Content
Merck
All Photos(2)

Key Documents

SAB4501643

Sigma-Aldrich

Anti-VEGFR2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

FLK1, KDR, kinase insert domain receptor, protein-tyrosine kinase receptor Flk-1, vascular endothelial growth factor receptor 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 151 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KDR(3791)

General description

Anti-VEGFR2 Antibody detects endogenous levels of total VEGFR2 protein.
The vascular endothelial growth factor receptor-2 (VEGFR-2) is also called as kinase insert domain containing receptor (KDR). There are seven immunoglobulin-like (Ig-like) domains at the  extracellular region (EC) of VEGFR-2. This gene is mapped to human chromosome 4q12.

Immunogen

The antiserum was produced against synthesized peptide derived from human VEGFR2.

Immunogen Range: 1141-1190

Biochem/physiol Actions

The immunoglobulin-like (Ig-like) domains of the vascular endothelial growth factor receptor-2 (VEGFR-2) are essential for specific ligand binding and receptor dimerization. It helps to activate multiple signaling cascades. VEGF-KDR participates in angiogenesis. VEGFR2 is involved in progression of several cancers like gliomas, malignant peripheral nerve sheath tumors, triple-negative breast cancer (TNBC) and gastrointestinal stromal tumors (GISTs).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erica W Mandell et al.
American journal of respiratory cell and molecular biology, 63(1), 79-91 (2020-03-07)
Vitamin D deficiency (VDD) during pregnancy is associated with increased respiratory morbidities and risk for chronic lung disease after preterm birth. However, the direct effects of maternal VDD on perinatal lung structure and function and whether maternal VDD increases the
Characterization of the amplicon on chromosomal segment 4q12 in glioblastoma multiforme.
Holtkamp N, et al.
Neuro-Oncology, 9(3), 291-297 (2007)
Vascular endothelial growth factor receptor-2 polymorphisms have protective effect against the development of tendinopathy in volleyball athletes.
Salles JI, et al.
PLoS ONE, 11(12), e0167717-e0167717 (2016)
Immunoglobulin-like domain 4-mediated ligand-independent dimerization triggers VEGFR-2 activation in HUVECs and VEGFR2-positive breast cancer cells.
Zhang S, et al.
Breast Cancer Research and Treatment, 163(3), 423-434 (2017)
The three receptor tyrosine kinases c-KIT, VEGFR2 and PDGFRα, closely spaced at 4q12, show increased protein expression in triple-negative breast cancer.
Jansson S, et al.
PLoS ONE, 9(7), e102176-e102176 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service