Skip to Content
Merck
All Photos(2)

Key Documents

SAB4501395

Sigma-Aldrich

Anti-Acetyl-CoA Carboxylase, N-Terminal antibody produced in rabbit

affinity isolated antibody

Synonym(s):

ACAC, ACACA, ACC-α, ACCA, Acetyl-CoA carboxylase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 265 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACACA(31)

Related Categories

General description

Anti-Acetyl-CoA Carboxylase Antibody detects endogenous levels of total Acetyl-CoA Carboxylase protein.
Acetyl-CoA carboxylase 1 (ACC1) is a cytosolic enzyme. It is located on human chromosome 17q12.

Immunogen

The antiserum was produced against synthesized peptide derived from human Acetyl-CoA Carboxylase.

Immunogen Range: 46-95

Biochem/physiol Actions

Acetyl-CoA carboxylase 1 (ACC1) catalyzes the rate-limiting step in de novo fatty acid biosynthesis. Acetyl-CoA is carboxylated to malonyl-CoA with the help of acetyl-CoA carboxylase (ACACA) in the fatty acid synthesis pathway. ACC1 is essential for the assembly of invadopodia.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Translational control of human acetyl-CoA carboxylase 1 mRNA is mediated by an internal ribosome entry site in response to ER stress, serum deprivation or hypoxia mimetic CoCl2
Damiano F, et al.
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1863(4), 388-398 (2018)
Metabolic regulation of invadopodia and invasion by acetyl-CoA carboxylase 1 and de novo lipogenesis
Scott K E N, et al.
PLoS ONE, 7(1), e29761-e29761 (2012)
Pietro Palumbo et al.
Gene, 538(2), 373-378 (2014-02-04)
Microdeletions of 17q12 including the hepatocyte nuclear factor 1 beta (HNF1B) gene, as well as point mutations of this gene, are associated with the Renal Cysts and Diabetes syndrome (RCAD, OMIM 137920) and genitourinary alterations. Also, microdeletions encompassing HNF1B were
Bruna Corominas-Faja et al.
Oncotarget, 5(18), 8306-8316 (2014-09-24)
Cancer stem cells (CSC) may take advantage of the Warburg effect-induced siphoning of metabolic intermediates into de novo fatty acid biosynthesis to increase self-renewal growth. We examined the anti-CSC effects of the antifungal polyketide soraphen A, a specific inhibitor of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service