Skip to Content
Merck
All Photos(2)

Documents

SAB4501074

Sigma-Aldrich

Anti-GAD1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

67 kDa glutamic acid decarboxylase, GAD-67, GAD1, Glutamate decarboxylase 1, Glutamate decarboxylase 67 kDa isoform

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 66 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GAD1(2571)

General description

Anti-GAD1 antibody detects endogenous levels of total GAD1 protein.
GAD1 gene (glutamate decarboxylase 1) codes for a glutamic acid decarboxylase enzyme (GAD67), that is of 67 kDa. It is mapped to human chromosome 2q31.1.

Immunogen

The antiserum was produced against synthesized peptide derived from human GAD1.

Immunogen Range: 471-520

Biochem/physiol Actions

GAD1 gene (glutamate decarboxylase 1) participates in GABA (γ-aminobutyric acid) synthesis. Variations in GAD1 results in hyperactivity in attention-deficit/hyperactivity disorder. It is also characterized with childhood-onset schizophrenia and cortical gray matter volume loss.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

GAD1 (2q31.1), which encodes glutamic acid decarboxylase (GAD67), is associated with childhood-onset schizophrenia and cortical gray matter volume loss.
Addington AM, et al.
Molecular Psychiatry, 10(6), 581-588 (2005)
GAD1 gene polymorphisms are associated with hyperactivity in Attention-Deficit/Hyperactivity Disorder
Bruxel EM, et al.
American Journal of Medical Genetics. Part B, Neuropsychiatric Genetics : the Official Publication of the International Society of Psychiatric Genetics, 171(8), 1099-1104 (2016)
GAD1 Gene Expression in Blood of Patients with First-Episode Psychosis
Yee JY, et al.
PLoS ONE, 12(1), e0170805-e0170805 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service