Skip to Content
Merck
All Photos(3)

Key Documents

SAB4500900

Sigma-Aldrich

Anti-LMTK2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Apoptosis-associated tyrosine kinase 2, Brain-enriched kinase, Kinase/phosphatase/inhibitor 2, Lemur tyrosine kinase 2, Serine/threonine protein kinase KPI-2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 164 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LMTK2(22853)

General description

Anti-LMTK2 antibody detects endogenous levels of total LMTK2 protein.
Lemur tyrosine kinase 2 (LMTK2) is a serine/threonine kinase, located in endosomal compartments. It is also termed as BREK (brain-enriched kinase), AATYK-2 (apoptosis-associated tyrosine kinase 2), cdk5, KPI-2 and Cprk. It is expressed at high levels in the plasma membrane. LMTK2 consists of 1503 amino acids and is located on human chromosome 7q21.

Immunogen

The antiserum was produced against synthesized peptide derived from human LMTK2.

Immunogen Range: 651-700

Application

Anti-LMTK2 antibody has been used in western blotting.

Biochem/physiol Actions

Lemur tyrosine kinase 2 (LMTK2) participates in endosomal membrane trafficking and neuronal physiology. It also participates in the differentiation of tissues and acts as a negative regulator of nerve growth factor (NGF)-induced neuronal differentiation.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rongjing Zhang et al.
Cancer letters, 454, 70-77 (2019-04-14)
Lemur tyrosine kinase 2 (LMTK2) belongs to both protein kinase and tyrosine kinase families. LMTK2 is less studied and little is known about its function. Here we demonstrate that LMTK2 modulates NF-κB activity and functions to promote colonic tumorigenesis. We
Signal dependent ER export of lemur tyrosine kinase 2
Butler EC and Bradbury NA
BMC Cell Biology, 16(1), 26-26 (2015)
Lemur tyrosine kinase 2 (LMTK2) is a determinant of cell sensitivity to apoptosis by regulating the levels of the BCL2 family members
Conti A, et al.
Cancer Letters, 389, 59-69 (2017)
Daniel F Cruz et al.
Frontiers in oncology, 11, 596861-596861 (2021-04-06)
Lemur tyrosine kinase 2 (LMTK2) is a transmembrane Ser/Thr kinase whose role has been increasingly recognized; however, when compared to other kinases, understanding of the LMTK2 networks and biological functions is still limited. Recent data have shown that transforming growth
Prostate cancer genomics: can we distinguish between indolent and fatal disease using genetic markers?
Wiklund F
Genome Medicine, 2(7), 45-45 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service