Skip to Content
Merck
All Photos(3)

Key Documents

SAB4500800

Sigma-Aldrich

Anti-AKT antibody produced in rabbit

affinity isolated antibody

Synonym(s):

AKT, AKT1 kinase, C-AKT, PKB, kinase Akt1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 55 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AKT1(207)

General description

Anti-AKT Antibody detects endogenous levels of total AKT protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human Akt.

Immunogen Range: 95-144

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Marroncini et al.
Journal of endocrinological investigation, 45(9), 1693-1708 (2022-05-24)
Hyponatremia, the most frequent electrolyte alteration in clinical practice, has been associated with a worse prognosis in cancer patients. On the other hand, a better outcome has been related to serum sodium normalization. In vitro studies have shown that low
Dibyendu Das et al.
Journal of food biochemistry, 46(12), e14385-e14385 (2022-09-08)
This study examined the antidiabetic efficacy of popular fermented soybean foods (FSF) of Northeast (NE) India. Results showed that among different FSF, aqueous extract of Hawaijar (AEH), a traditional FSF of Manipur, NE India, significantly augmented glucose utilization in cultured
Maryam Farzaneh Behelgardi et al.
Molecular biology reports (2022-02-11)
There is ample evidence to suggest that vascular endothelial growth factor (VEGF) is a potent mitogen factor in vasculogenesis and angiogenesis and that blockade of VEGF-mediated signals can also prevent tumor growth via enforcing cell apoptosis. In the current study
Giada Marroncini et al.
Molecular and cellular endocrinology, 537, 111419-111419 (2021-08-15)
Hyponatraemia is frequently observed in cancer patients and can be due to the syndrome of inappropriate anti-diuresis (SIAD), related to ectopic vasopressin secretion, particularly in small cell lung cancer (SCLC). Hyponatraemia is associated with a worse outcome in cancer patients.
Anjum Dihingia et al.
Annals of the New York Academy of Sciences, 1462(1), 104-117 (2019-10-05)
The role of gamma-glutamyl-carboxylated growth arrest-specific 6 (cGas6) in mediating the beneficial effect of vitamin K (VK) on regulating glucose metabolism remains elusive. We took a three-pronged approach-evaluating human type 2 diabetes (T2D), high-fat diet (HFD)-fed mice, and in vitro

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service