Skip to Content
Merck
All Photos(1)

Documents

SAB4301399

Sigma-Aldrich

Anti-phospho-DRP1 (pSer637) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

DLP1, DNM1L, DRP1, DVLP, Dymple

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

82 kDa

species reactivity

human

concentration

1.0 mg/mL

technique(s)

western blot: 1:500-1:1000 (Cell Lysate)

isotype

IgG

accession no.

NP_036192.2.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer637)

Gene Information

human ... DNM1L(10059)

General description

Dynamin related protein 1 (DRP1) is a dynamin-related guanosine triphosphatase. The gene is located on human chromosome 12p11.21. DRP1 is a cytosolic protein, which is also present on the outer mitochondrial membrane.

Specificity

The antibody detects endogenous levels of DRP1 only when phosphorylated at serine 637.

Immunogen

Peptide sequence around phosphorylation site of Serine637(K-L-S(p)-A-R) derived from Human DRP1.

Application

Anti-phospho-DRP1 (pSer637) antibody produced in rabbit has been used in immunoblotting and western blotting.

Biochem/physiol Actions

Dynamin related protein 1 (DRP1) is an important regulator of mitochondrial fission and peroxisomal division. It regulates proper mitochondrial function, apoptosis and necrosis. Mutations in this gene is associated with Alzheimer′s disease.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

UBIAD1 protects against oxygen-glucose deprivation/reperfusion-induced multiple subcellular organelles injury through PI3K/AKT pathway in N2A cells
Huang Y and Hu Z
Journal of Cellular Physiology, 233(9), 7480-7496 (2018)
Legionella pneumophila modulates mitochondrial dynamics to trigger metabolic repurposing of infected macrophages
Escoll P, et al.
Cell host & microbe, 22(3), 302-316 (2017)
6-Hydroxydopamine (6-OHDA) induces Drp1-dependent mitochondrial fragmentation in SH-SY5Y cells
Gomez-Lazaro, et al.
Free Radical Biology & Medicine, 44(11), 1960-1969 (2008)
Downregulation of Drp1, a fission regulator, is associated with human lung and colon cancers
Kim YY, et al.
Acta biochimica et biophysica Sinica, 50(2), 209-215 (2018)
S-nitrosylation of Drp1 mediates ?-amyloid-related mitochondrial fission and neuronal injury
Cho DH, et al.
Science, 324(5923), 102-105 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service