Skip to Content
Merck
All Photos(2)

Key Documents

SAB4300213

Sigma-Aldrich

Anti-phospho-H2AFX (pSer139) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-H2A histone family, member X antibody produced in rabbit, Anti-H2A.X antibody produced in rabbit, Anti-H2A/X antibody produced in rabbit, Anti-H2AX antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~15 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

indirect immunofluorescence: 1:100-1:200
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(Q-A-SP-Q-E)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer139)

Gene Information

human ... H2AFX(3014)

Related Categories

Immunogen

Peptide sequence around phosphorylation site of serine 139 (Q-A-S(p)-Q-E), according to the protein H2AFX.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Variant histone H2A which replaces conventional H2A in a subset of nucleosomes. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling. Required for checkpoint-mediated arrest of cell cycle progression in response to low doses of ionizing radiation and for efficient repair of DNA double strand breaks (DSBs) specifically when modified by C-terminal phosphorylation.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Konstantin Byrgazov et al.
Therapeutic advances in medical oncology, 12, 1758835920937891-1758835920937891 (2020-08-11)
Low survival rates in metastatic high-grade osteosarcoma (HGOS) have remained stagnant for the last three decades. This study aims to investigate the role of aminopeptidase N (ANPEP) in HGOS progression and its targeting with a novel lipophilic peptidase-enhanced cytotoxic compound
Sumedh S Shah et al.
Cancers, 11(2) (2019-02-03)
Glioblastoma (GBM) is the most common and aggressive primary brain tumor in adults, and despite optimized treatment options, median survival remains dismal. Contemporary evidence suggests disease recurrence results from expansion of a robustly radioresistant subset of GBM progenitor cells, termed
Jordan Guillon et al.
Cell death & disease, 10(3), 199-199 (2019-03-01)
Senescence is a tumor-suppressive mechanism induced by telomere shortening, oncogenes, or chemotherapy treatment. Although it is clear that this suppressive pathway leads to a permanent arrest in primary cells, this might not be the case in cancer cells that have
Ding Wu et al.
BMC genomics, 20(1), 290-290 (2019-04-17)
Diverse stresses including genotoxic therapy can induce proliferating cancer cells to undergo cellular senescence and take on the characteristic phenotypes of replicative cellular aging. This accelerated or therapy-induced senescence has been alternatively proposed to contribute to therapeutic efficacy or resistance.
Yixuan Zhang et al.
Gene, 711, 143949-143949 (2019-07-01)
As a transcriptional repressor, Chromobox 8 (CBX8) overexpression is found to be associated with tumorigenesis in several cancers. However, its role in radiotherapy resistance remains poorly characterized. Our study is the first to explore the correlation between CBX8 and radioresistance.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service