Skip to Content
Merck
All Photos(3)

Key Documents

SAB4200810

Sigma-Aldrich

Anti-Podocin, Mouse monoclonal

clone PD-65, purified from hybridoma cell culture

Synonym(s):

NPHS2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

PD-65, monoclonal

form

buffered aqueous solution

mol wt

~42 kDa

species reactivity

bovine, rat, human, mouse

packaging

antibody small pack of 25 μL

concentration

1 mg/mL

technique(s)

immunoblotting: 0.25-0.5 μg/mL using using human HEK-293T cells.
immunofluorescence: 1-2 μg/mL using human HEK-293T cells.

isotype

IgG2a

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NPHS2(7827)

General description

Podocytes are highly specialized visceral epithelial cells that cover the outer aspect of the glomerulus in the kidney. The foot processes of adjacent podocytes regularly interdigitate leaving filtration slits that are bridged by a thin slit diaphragm. This is an extracellular adherens-like junction that serves as a barrier to macromolecules and plays a crucial role in the glomerular filtration process. Podocin (NPHS2 protein) is an important member of a group of proteins shown to be associated with the slit diaphragm. Podocin belongs to the band-7-stomatin family of lipid raft-associated proteins. It is a hairpin-like integral membrane protein with intracellular N- and C- termini. Podocin is located at the insertion site of the slit membrane, and is thought to act as a scaffold protein required to maintain or regulate the structural integrity of the slit diaphragm. It interacts there with nephrin (NPHS1 protein), a protein critical in development and function of the kidney filtration barrier and the adapter protein CD2-AP. Podocin is anchored in the glycosphingolipids and cholesterol rich lipid rafts of the outer leaflet of the plasma membrane and is partially localized with the actin cytoskeleton. In vitro studies suggest its involvement in nephrin signaling facilitation via AP-1 in HEK cells. In addition to its presence in the kidney, it was also reported in rat nervous tissue and possibly in vascular smooth muscle.

Immunogen

Synthetic peptide of human Podocin , conjugated to KLH

Application

The antibody may be used in various immunochemical techniques including Immunobloting (~42kDa) and Immunofluorescence. Detection of the Podocin band by Immunoblotting is specifically inhibited by the immunogen.a

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative

Other Notes

This product is for R&D use only, not for drug, household, or other uses.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinwang Zhu et al.
International journal of molecular medicine, 53(3) (2024-01-19)
The senolytics dasatinib and quercetin (DQ) alleviate age‑related disorders. However, limited information is available regarding the effects of DQ on diabetic kidney disease (DKD). The present study aimed to explore the effects of DQ on DKD and its potential molecular
Wang Aixue et al.
Skin research and technology : official journal of International Society for Bioengineering and the Skin (ISBS) [and] International Society for Digital Imaging of Skin (ISDIS) [and] International Society for Skin Imaging (ISSI), 30(2), e13562-e13562 (2024-01-27)
Pathological studies have shown an association between psoriasis and renal podocyte injury, and the specific mechanism of podocyte injury in psoriasis remains unclear, with no effective treatments currently available. This study aimed to investigate the underlying mechanisms of podocyte and
Hua Sun et al.
Kidney360, 4(2), 162-176 (2023-02-24)
The expression of dynein is increased in human and rodent models of diabetic nephropathy (DN), eliciting a new dynein-driven pathogenesis. Uncontrolled dynein impairs the molecular sieve of kidney by remodeling the postendocytic triage and homeostasis of nephrin. The delineation of
Francesco De Luca et al.
PloS one, 18(1), e0280270-e0280270 (2023-01-18)
The podocyte is a specialized cell type critically involved in maintaining the selective filtration barrier of the kidney. Podocytes are primary or secondary targets for a multitude of kidney diseases. Despite intense investigation, the transcriptome and proteome of human podocytes
Hang Mei et al.
International journal of biological sciences, 20(9), 3317-3333 (2024-07-12)
The glomerular podocyte, a terminally differentiated cell, is crucial for the integrity of the glomerular filtration barrier. The re-entry of podocytes into the mitotic phase results in injuries or death, known as mitotic catastrophe (MC), which significantly contributes to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service