Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200804

Sigma-Aldrich

Anti-Goat/sheep IgG antibody, Mouse monoclonal

clone GT-34, purified from hybridoma cell culture

Synonym(s):

Goat/Sheep immunoglobulin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

GT-34, monoclonal

form

buffered aqueous solution

species reactivity

goat, sheep

packaging

antibody small pack of 25 μL

concentration

~1 mg/mL

technique(s)

indirect ELISA: 0.25-0.5 μg/mL using 5μg/ml Goat IgG for coating.

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Goat and sheep antibodies are widely used as primary antibodies against many antigens in various assay techniques in research and clinical practices. Polyclonal second antibodies to goat and sheep may suffer from a lack of unique specificity to goat or sheep IgGs, and may cross react with other IgGs from human, mouse, rat or rabbit origin. As a result, an extensive adsorbing stage has to be integrated in the manufacturing process of most polyclonal antibodies. The use of a monoclonal antibody specific to goat or sheep IgG which is devoid of any binding capacity to human and many other species, can therefore serve as an essential tool and a useful control in many tests and applications.
Monoclonal Anti-Goat/sheep IgG (mouse IgG1 isotype) is derived from the GT-34 hybridoma, produced by the fusion of mouse myeloma cells and splenocytes from a mouse immunized with purified goat immunoglobulins (IgGs). Immunoglobulin G (IgG) belongs to the immunoglobulin family and is a widely expressed serum antibody. An immunoglobulin has two heavy chain and two light chain connected by disulfide bond. It mainly helps in immune defense. It is a glycoprotein. IgG is a major class of immunoglobulin. Goat IgG has two subclasses- IgG1 and IgG2.

Specificity

Monoclonal Anti-Goat/sheep recognizes an epitope located on the heavy chain of goat IgG1 and IgG2. The antibody shows strong cross-reactivity with sheep IgG and weak cross-reactivity with bovine IgG. No cross-reactivity is observed with IgG derived from human, monkey, guinea pig, rat, horse, fish, donkey, dog, chicken, pig, mouse, rabbit or cat origin. The antibody is recommended to use in various immunological techniques, including ELISA, Immunoblotting, immunofluorescence and Immunohistochemistry.

Immunogen

purified goat immunoglobulins (IgGs).

Biochem/physiol Actions

Immunoglobulin G (IgG) participates in hypersensitivity type II and type III.

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative.

Other Notes

This product is for R&D use only, not for drug, household, or other uses.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Specific IgG for cat allergens in patients with allergic conjunctivitis.
Miyama A, et al.
International Ophthalmology, 35(4), 575-586 (2015)
Antibody structure, instability, and formulation.
Wang W
Journal of Pharmaceutical Sciences, 96(1), 1-26 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service