Skip to Content
Merck
All Photos(2)

Key Documents

SAB4200783

Sigma-Aldrich

Anti-Human IgG2 antibody, Mouse monoclonal

clone HP-6014, purified from hybridoma cell culture

Synonym(s):

Anti-Human immunoglobulin G2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

Quality Level

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

HP-6014, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

immunofluorescence: suitable
indirect ELISA: 0.07-0.15 μg/mL using 1 μg/mL Human IgG2 myeloma for coating

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Anti-Human IgG2 antibody, Mouse monoclonal (mouse IgG1 isotype) is derived from the HP-6014 hybridoma, produced by the fusion of mouse myeloma cells and splenocytes from a mouse immunized with purified human IgG2 myeloma proteins covalently coupled to polyaminostyrene (PAS) microbeads. Human IgG consist of four subclasses (1-4) that can be recognized by antigenic differences in their heavy chains. They constitute approximately 65, 30, 5, and 4% of the total IgG, respectively.

Immunogen

Purified human IgG2 myeloma protein

Application

Anti-Human IgG2 antibody, Mouse monoclonal may be used in immunological techniques, including enzyme linked immunosorbent assay (ELISA), immunofluorescence, hemagglutination (HA) and hemagglutination inhibition.

Biochem/physiol Actions

Lipopolysaccharides stimulate an IgG2 response in peripheral blood leukocytes (PBL) and an IgG1 response in the spleen. Only IgG1 and IgG3 are capable of adherence to mononuclear phagocytes while IgG2 and IgG4 autoantibodies are much less efficient. The amount of the different IgG subclasses present in the blood shows variation with age. For example, IgG1 and IgG3 reach normal adult levels by 5-7 years of age while IgG2 and IgG4 levels raise more slowly, reaching adult levels at about 10 years of age. Serum IgG subclass deficiencies have been recorded for different patient groups. For example, IgG2 and IgG4 deficiency is found in patients of ataxia telangiectasia. Low IgG2 levels were found in patients with Systemic lupus erythematosus (SLE) and juvenile diabetes mellitus.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Age-associated distribution of normal B-cell and plasma cell subsets in peripheral blood
Blanco E, et al.
The Journal of Allergy and Clinical Immunology, 141(6), 2208-2219 (2018)
Immunoglobulin G; structure and functional implications of different subclass modifications in initiation and resolution of allergy
Scott-Taylor TH, et al.
Immunity, inflammation and disease, 6, 13-33 (2018)
Effect of anti-epileptic drugs on serum level of IgG subclasses
Ashrafi MR, et al.
Iranian journal of pediatrics, 20(3), 269-269 (2010)
Immunodeficiency and infections in ataxia-telangiectasia
Nowak-Wegrzyn A, et al.
The Journal of Pediatrics, 144(4), 505-511 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service