Skip to Content
Merck
All Photos(2)

Key Documents

SAB4200304

Sigma-Aldrich

Anti-Atg7 antibody, Mouse monoclonal

clone ATG7-13, purified from hybridoma cell culture

Synonym(s):

Anti-ATG12-activating enzyme E1 ATG7, Anti-Autophagy related 7 homolog (S. cerevisiae), APG7-like, APG7L, GSA7, APG7 autophagy 7-like, Anti-Autophagy-related protein 7, Anti-HAGP7, Anti-Ubiquitin activating enzyme E1-like protein, Anti-Ubiquitin-activating enzyme E1-like protein, Anti-Ubiquitin-like modifier-activating enzyme ATG7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

ATG7-13, monoclonal

form

buffered aqueous solution

mol wt

antigen ~75 kDa

species reactivity

human, mouse, rat

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): suitable
western blot: 2-4 μg/mL using whole extracts of HEK-293T or U87 cells

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATG7(10533)
mouse ... Atg7(74244)
rat ... Atg7(312647)

General description

Autophagy related 7 (ATG7) is encoded by the gene mapped to human chromosome 3p25.3. The encoded protein has ubiquitous expression.
Monoclonal Anti-Atg7 (mouse IgG1 isotype) is derived from the hybridoma ATG7-13 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide corresponding to a sequence located near the N-terminus of human Atg7.

Immunogen

synthetic peptide corresponding to a sequence located near the N-terminus of human Atg7, conjugated to KLH. The corresponding sequence is identical in mouse and rat Atg7.

Application

Monoclonal Anti-Atg7 antibody produced in mouse has been used in:
  • immunofluorescence
  • immunohistochemistry (IHC)
  • western blotting
  • immunoprecipitation

Biochem/physiol Actions

Atg7 can activate all three Atg8 mammalian homologues, (Golgi-associated ATPase enhancer of 16 kDa (GATE-16), GABA receptor-associated protein (GABARAP) and microtubule-associated protein light chain 3 (MAP-LC3). It forms a homodimer through the C-terminal region that is important for enzyme substrate interaction and E1-E2 complex formation. Atg7 was found to be essential for amino acid supply in neonates and starvation− induced bulk degradation of proteins and organelles in mice. Loss of Atg7 results in the accumulation of abnormal organelles and ubiquitin positive aggregates and leads to neurodegeneration.
Autophagy related 7 (ATG7) plays a vital role in the regulation of adult hematopoietic stem cells (HSCs). ATG7 expression is elevated in eosinophilic esophagitis (EoE). Hence, this can be used as a potent tissue biomarker for diagnosis of EoE inflammation in pediatric patients. The encoded protein plays an essential role in autophagy vesicle elongation. In addition, it is also implicated in two vital ubiquitin-like reactions such as LC3 lipidation and Atg5/12 conjugation. Down regulated expression of Atg7 increases the sensitivity to chemotherapy and disables stroma-mediated chemoresistance in acute myeloid leukemia (AML).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Autophagy protects cardiomyocytes from the myocardial ischaemia-reperfusion injury through the clearance of CLP36
Li S, et al.
Open Biology, 6(8), 160177-160177 (2016)
The autophagy protein Atg7 is essential for hematopoietic stem cell maintenance
Mortensen M
The Journal of Experimental Medicine, 208, 455-467 (2011)
In vivo autophagy and biogenesis of autophagosomes within male haploid cells during spermiogenesis
Yang P, et al.
Oncotarget, 8(34), 56791-56791 (2017)
Atg7 suppression enhances chemotherapeutic agent sensitivity and overcomes stroma-mediated chemoresistance in acute myeloid leukemia
Piya S, et.al.
Blood, 128, 1260-1269 (2016)
Atg7 is required for acrosome biogenesis during spermatogenesis in mice
Wang H, et al.
Cell Research, 24(7), 852-852 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service