Skip to Content
Merck
All Photos(3)

Key Documents

SAB3500390

Sigma-Aldrich

Anti-ELOVL7 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Elongation of very long chain fatty acids protein 7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ELOVL7(79993)

General description

ELOVL (Elongation of very long chain fatty acids) fatty acid elongase 7 is expressed in most tissues except heart and skeletal muscles. In human chromosome, the gene ELOVL7 is localised on 5q12.1.

Immunogen

ELOVL7 antibody was raised against a 15 amino acid peptide near the amino terminus of human ELOVL7.

Biochem/physiol Actions

ELOVL7 catalyses the elongation of long chain fatty acids, preferably saturated long chain fatty acids. Elovl7 exhibited activity towards C16-C20 acyl coenzyme A with maximum affinity towards carbon length of C18. Elovl7 expression is regulated by sterol regulatory element-binding protein 1 (SREBP1) and also by other lipogenic enzymes. The expression levels of Elovl7 is higher in prostate cancer leading to its progression.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500390.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cockayne syndrome caused by paternally inherited 5Mb deletion of 10q11. 2 and a frameshift mutation of ERCC6
Ghai SJ, et al.
European Journal of Medical Genetics, 54(3), 272-276 (2011)
Yuecheng Xi et al.
Journal of virology, 93(21) (2019-08-09)
Human cytomegalovirus (HCMV) replication requires host metabolism. Infection alters the activity in multiple metabolic pathways, including increasing fatty acid elongation and lipid synthesis. The virus-host interactions regulating the metabolic changes associated with replication are essential for infection. While multiple host
Yuecheng Xi et al.
mBio, 12(3) (2021-05-06)
Stress and virus infection regulate lipid metabolism. Human cytomegalovirus (HCMV) infection induces fatty acid (FA) elongation and increases the abundance of lipids with very-long-chain FA (VLCFA) tails. While reprogramming of metabolism can be stress related, the role of stress in
Tatsuro Naganuma et al.
FEBS letters, 585(20), 3337-3341 (2011-10-01)
Very long-chain fatty acids (VLCFAs) have a variety of physiological functions and are related to numerous disorders. The key step of VLCFA elongation is catalyzed by members of the elongase family, ELOVLs. Mammals have seven ELOVLs (ELOVL1-7), yet none of
Yusuke Ohno et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(43), 18439-18444 (2010-10-13)
Very long-chain fatty acids (VLCFAs) exert a variety of cellular functions and are associated with numerous diseases. However, the precise pathway behind their elongation has remained elusive. Moreover, few regulatory mechanisms for VLCFAs synthesis have been identified. Elongases catalyze the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service