Skip to Content
Merck
All Photos(1)

Key Documents

SAB2900383

Sigma-Aldrich

Anti-SFRP1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

frza, sarp2, secreted frizzled related protein, sfrp-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect immunofluorescence: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SFRP1(6422)

Related Categories

General description

SFRP1 (secreted frizzled-related protein 1) is a 35.4 kDa antagonist protein.
The encoded protein by SFRP1 gene has a cysteine-rich domain similar to the WNT-binding site of Frizzled receptors. It belongs to the WNT pathway. This gene is located on human chromosome 8p12–11.1.
The gene SFRP1 (secreted frizzled related protein 1) is mapped to human chromosome 8p22–p12, a region frequently deleted in breast cancer. It is highly expressed in normal breast tissue.

Immunogen

Synthetic peptide from human SFRP1.

Application

Anti-SFRP1 antibody has been used in western blot analysis.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

SFRP1 (secreted frizzled-related protein 1) plays a major role in the progression of several malignancies. It also plays a vital role in antagonizing Wnt signaling. Overexpression of SFRP1 is linked with apoptosis.
Secreted Frizzled-related protein 1 (SFRP1) is a secreted inhibitor of Wnt signaling pathway. DNA damage or oxidative stress causes cellular senescence, which leads to the overexpression of sfrp1. This protein induces senescence phenotypes via the inactivation of Wnt signaling and activation of the retinoblastoma (Rb) pathway. Mutations or aberrant methylation in this gene impair the induction of senescence in several cancers, including breast cancer, bladder cancer, and colorectal tumorigenesis.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.02 M potassium phosphate, 0.15 M sodium chloride, pH 7.2, 0.01% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deletions of chromosome 8p and loss of sFRP1 expression are progression markers of papillary bladder cancer
Stoehr R, et al.
Laboratory Investigation; a Journal of Technical Methods and Pathology, 84(4), 465-465 (2004)
Effects of supplementation with nondigestible carbohydrates on fecal calprotectin and on epigenetic regulation of SFRP1 expression in the large-bowel mucosa of healthy individuals
Malcomson FC, et al.
American Journal of Clinical Nutrition, 105(2), 400-410 (2017)
WNT pathway and mammary carcinogenesis: Loss of expression of candidate tumor suppressor gene SFRP1 in most invasive carcinomas except of the medullary type
Ugolini F, et al.
Oncogene, 20(41), 5810-5810 (2001)
Influence of secreted frizzled receptor protein 1 (SFRP1) on neoadjuvant chemotherapy in triple negative breast cancer does not rely on WNT signaling
Bernemann C, et al.
Molecular Cancer, 13(1), 174-174 (2014)
Xiaobin Wang et al.
Oncology letters, 4(2), 334-338 (2012-07-31)
The aims of this study were to determine the methylation and expression status of secreted Frizzled-related protein 1 (SFRP1) in bladder cancer, to explore the mechanisms involved and to study the role of SFRP1 in the pathogenesis of bladder cancer.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service