Skip to Content
Merck
All Photos(9)

Key Documents

SAB2702167

Sigma-Aldrich

Monoclonal Anti-TSG101 antibody produced in mouse

Synonym(s):

TSG10, VPS23

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA
FACS
ICC
IF
IHC
IP
WB
clone:
4A10
monoclonal
species reactivity:
human, monkey, mouse, rat, hamster
citations:
11
technique(s):
ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: 1:500-1:3000

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4A10
monoclonal

form

liquid

mol wt

46 kDa

species reactivity

human, monkey, mouse, rat, hamster

concentration

1 mg/mL

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: 1:500-1:3000

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TSG101(7251)

General description

Tumor susceptibility gene 101 (TSG101) is an endosomal sorting complex required for transport (ESCRT) component, present on endosomal membranes. This gene is located on chromosome 11p15. The TSG101 gene encodes a 46 kDa protein and contains a proline-rich domain and a leucine-zipper motif within a coiled-coil domain near the carboxyl terminus.

Immunogen

Amino acids 167-374 of TSG101 protein expressed in E. coli.

Application

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Suggested fixation for ICC/IF: 3-3.5% PFA with 0.05% saponin treatment. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Monoclonal Anti-TSG101 antibody has been used in western blotting and immunoprecipitation (IP) analyses.

Biochem/physiol Actions

The TSG101 gene encodes a 46 kDa protein and contains a proline-rich domain and a luecine-zipper motif within a coiled-coil domain near the carboxyl terminus.
Tumor susceptibility gene 101 (TSG101) is essential for the generation of central supramolecular activation cluster, and the signal termination of T cell receptor (TCR) microclusters (MC). It plays an important role in TCR downregulation and separation of TCR- major histocompatibility complex (MHC)-peptide from signaling complexes, that are enriched with protein kinase C θ (PKCθ).
TSG101-4A10 recognizes TSG101 protein, the product of a recently identified tumor susceptibility gene whose inactivation in mouse fibroblasts results in cell transformation and the ability of those cells to form tumors in nude mice.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Phosphate-buffered saline, no preservative added.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bone marrow niche trafficking of miR-126 controls the self-renewal of leukemia stem cells in chronic myelogenous leukemia.
Zhang B, et al.
Nature Medicine, 24(4), 450-450 (2018)
Bettina Mannerström et al.
Epigenetics, 14(4), 352-364 (2019-03-26)
Extracellular vesicles (EVs) are central to intercellular communication and play an important role in cancer progression and development. Osteosarcoma (OS) is an aggressive bone tumour, characterized by the presence of malignant mesenchymal cells. The specific tumour-driving genetic alterations that are
Absence of TSG101 transcript abnormalities in human cancers.
Trink B, et al.
Oncogene, 16(21), 2815-2815 (1998)
Shengqiang Xu et al.
ACS biomaterials science & engineering, 4(2), 654-662 (2018-04-07)
MicroRNA (miRNA) in urine has been considered as a potential biomarker for early-stage diagnosis of multiple diseases like urinary system cancer, kidney injury and diabetes, owing to their many demonstrated advantages including long-term stability and noninvasiveness. However, the traditional enrichment
Essential role of ubiquitin and TSG101 protein in formation and function of the central supramolecular activation cluster.
Vardhana S, et al.
Immunity, 32(4), 531-540 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service