Skip to Content
Merck
All Photos(13)

Key Documents

SAB2700282

Sigma-Aldrich

Anti-GPC1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

FLJ38078, GPC1, glypican

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human

technique(s)

ELISA: suitable
flow cytometry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: 500-3000

NCBI accession no.

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GPC1(2817)

General description

Glypican 1 (GPC1) is located at 2q37.3 on the human chromosome. It belongs to the family of glycosylphosphatidylinositol-anchored heparan sulfate proteoglycan. GPC1 is localized in the vascular system. GPC1 consists of an N-terminal secretory signal peptide, 14 conserved cysteine residues, a heparan sulfate (HS) attachment domain near the C terminus and a hydrophobic domain for attachment of the glycosylphosphatidylinositol (GPI) at the C terminus.

Immunogen

Recombinant protein encompassing a sequence within the center region of human Glypican 1.

Application

Anti-GPC1 antibody produced in rabbit has been used in flow cytometry and immunostaining.

Biochem/physiol Actions

Glypican 1 (GPC1) plays an important role in cell growth, differentiation and morphogenesis. It also plays a role in cell migration, adhesion, anti-coagulation, lipoprotein metabolism and modulation of growth factors. GPC1 exhibits chaperone-like activity by restoring the binding activity of oxidized VEGF165 (vascular endothelial growth factors-165). Overexpression of GPC1 results in over accumulation of β-amyloid protein (Aβ) in the Alzheimer′s disease (AD) brain. Mutation in this gene is associated with Niemann-Pick C1 disease.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

1XPBS, 20% Glycerol (pH7). 0.025% ProClin 300 was added as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glypican-1 as an Abeta binding HSPG in the human brain: Its localization in DIG domains and possible roles in the pathogenesis of Alzheimer?s disease
Watanabe N, et al.
Faseb Journal, 18(9), 1013-1015 (2004)
Characterization of Slit protein interactions with glypican-1
Ronca F, et al.
Test, 276(31), 29141-29147 (2001)
Nelson S Yee et al.
Biomedicines, 8(12) (2020-12-11)
Pancreatic carcinoma (PC) is highly metastatic, and it tends to be detected at advanced stages. Identifying and developing biomarkers for early detection of PC is crucial for a potentially curative treatment. Extracellular vesicles (EVs) are bilayer lipid membrane-structured nanovesicles found
Aikaterini Emmanouilidi et al.
Proteomics, 19(8), e1800158-e1800158 (2019-03-21)
Exosomes are small extracellular membrane vesicles important in intercellular communication, with their oncogenic cargo attributed to tumor progression and pre-metastatic niche formation. To gain an insight into key differences in oncogenic composition of exosomes, human non-malignant epithelial and pancreatic cancer
Glypican-1 is a VEGF165 binding proteoglycan that acts as an extracellular chaperone for VEGF165
Gengrinovitch S, et al.
Test, 274(16), 10816-10822 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service