Skip to Content
Merck
All Photos(8)

Key Documents

SAB2700181

Sigma-Aldrich

Anti-NS3 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Dengue virus 2 Nonstructural protein NS3, Non-structure protein 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

Dengue virus

technique(s)

flow cytometry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: 500-3000

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

General description

Non-structure protein 3 (NS3), also known as dengue virus type 2 (DEN2), belongs to the Flaviviridae family. NS3 is a multifunctional protein with a molecular mass of about 69 kDa. It contains serine protease domain (NS3-pro) and it needs the hydrophilic domain of NS2B (NS2BH) for activation.

Immunogen

Recombinant protein fragment contain a sequence corresponding to a region within amino acids 354 and 568 of NS3 (Dengue virus 2) according to NP_739587

Application

Suggested starting dilutions are as follows: FACS: Assay-dependent dilution, ICC/IF: 1:100-1:2000, IHC: Assay-dependent dilution, IHC-P: Assay-dependent dilution, WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Anti-NS3 antibody produced in rabbit has been used in flow cytometry.

Biochem/physiol Actions

Non-structure protein 3 (NS3) along with NS2B play a vital role in proteolytic cleavage of dengue virus (DEN) non-structural proteins. NS3 also acts as a RNA-stimulated nucleoside-triphosphatase (NTPase)/RNA helicase and a 5′-RNA triphosphatase (RTPase).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

1XPBS, 20% Glycerol (pH7). 0.025% ProClin 300 was added as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

RIG-I-like receptor activation by dengue virus drives follicular T helper cell formation and antibody production.
Sprokholt JK, et al.
PLoS Pathogens, 13(11), e1006738-e1006738 (2017)
Alexandra P M Cloherty et al.
Frontiers in immunology, 15, 1260439-1260439 (2024-06-12)
Dengue virus (DENV), transmitted by infected mosquitoes, is a major public health concern, with approximately half the world's population at risk for infection. Recent decades have increasing incidence of dengue-associated disease alongside growing frequency of outbreaks. Although promising progress has
Both nonstructural proteins NS2B and NS3 are required for the proteolytic processing of dengue virus nonstructural proteins.
Falgout B, et al.
Journal of Virology, 65(5), 2467-2475 (1991)
Joris K Sprokholt et al.
Journal of immunology (Baltimore, Md. : 1950), 198(12), 4764-4771 (2017-05-17)
Dengue virus (DENV) causes 400 million infections annually and is one of several viruses that can cause viral hemorrhagic fever, which is characterized by uncontrolled immune activation resulting in high fever and internal bleeding. Although the underlying mechanisms are unknown
Modulation of the nucleoside triphosphatase/RNA helicase and 5′-RNA triphosphatase activities of Dengue virus type 2 nonstructural protein 3 (NS3) by interaction with NS5, the RNA-dependent RNA polymerase.
Yon C, et al.
The Journal of Biological Chemistry (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service