Skip to Content
Merck
All Photos(4)

Key Documents

SAB2108079

Sigma-Aldrich

Anti-MAGEA4 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

35kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunoblotting: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MAGEA4(4103)

Immunogen

Synthetic peptide directed towards the C terminal region of human MAGEA4

Application

Anti-MAGEA4 antibody produced in rabbit is suitable for immunoblotting techniques.

Biochem/physiol Actions

MAGEA4 is a member of the MAGEA family. The members of this family are proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita.This gene is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita. At least four variants encoding the same protein have been found for this gene.

Sequence

Synthetic peptide located within the following region: ENYLEYRQVPGSNPARYEFLWGPRALAETSYVKVLEHVVRVNARVRIAYP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sheetal Bhan et al.
Oncology reports, 28(4), 1498-1502 (2012-07-31)
Cancer testis antigens (CTAs) are proteins that are normally expressed only in male germ cells and are aberrantly upregulated in a variety of cancers such as melanomas and lung cancer. MAGEA proteins belong to Class I CTAs and are being
Yousri M Hussein et al.
Medical oncology (Northwood, London, England), 29(2), 994-999 (2011-04-01)
The dissemination of hepatocellular carcinoma (HCC) cells into the circulation plays a critical role in post-operative recurrence and metastasis. Early detection of metastatic tumor cells is critical to identify HCC patients at high risk of relapse. MAGE-3 and -4 genes
R C Hillig et al.
Journal of molecular biology, 310(5), 1167-1176 (2001-08-15)
The heterotrimeric complex of the human major histocompatibity complex (MHC) molecule HLA-A*0201, beta2-microglobulin and the decameric peptide GVYDGREHTV derived from the melanoma antigen (MAGE-A4 protein has been determined by X-ray crystallography at 1.4 A resolution. MAGE-A4 belongs to a family
M T Duffour et al.
European journal of immunology, 29(10), 3329-3337 (1999-10-30)
The MAGE-encoded antigens that are recognized by cytolytic T lymphocytes (CTL) are shared by many tumors and are strictly tumor specific. Clinical trials involving therapeutic vaccination of cancer patients with MAGE antigenic peptides or proteins are in progress. To increase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service