Skip to Content
Merck
All Photos(3)

Key Documents

SAB1411206

Sigma-Aldrich

Anti-MRPL12 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

5c5-2, FLJ60124, L12mt, MGC8610, MRP-L31/34, MRPL7, MRPL7/L12, RPML12

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 21.3 kDa

species reactivity

human

technique(s)

immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MRPL12(6182)

Related Categories

General description

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein which forms homodimers. In prokaryotic ribosomes, two L7/L12 dimers and one L10 protein form the L8 protein complex. (provided by RefSeq)

Immunogen

MRPL12 (NP_002940.2, 1 a.a. ~ 198 a.a) full-length human protein.

Sequence
MLPAAARPLWGPCLGLRAAAFRLARRQVPCVCAVRHMRSSGHQRCEALAGAPLDNAPKEYPPKIQQLVQDIASLTLLEISDLNELLKKTLKIQDVGLVPMGGVMSGAVPAAAAQEAVEEDIPIAKERTHFTVRLTEAKPVDKVKLIKEIKNYIQGINLVQAKKLVESLPQEIKANVAKAEAEKIKAALEAVGGTVVLE

Biochem/physiol Actions

MRPL12 (mitochondrial ribosomal protein) is involved in the mitochondrial translation. It directly interacts with the mitochondrial RNA polymerase (POLRMT) and triggers the mitochondrial transcriptional process. Research shows that the interaction between MRPL12 and POLRMT indicates its participation in the regulatory mechanism that coordinates mitochondrial transcription with translation and/or ribosome biogenesis during human mitochondrial gene expression. Study shows that MRPL12 may be involved in the differentiation of colonic epithelial cells and its reduced mitochondrial activity have been shown to be linked with tumor formation. It has been reported that mutated expression of MRPL12 associates with the growth retardation, neurological deterioration and mitochondrial translation deficiency.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhibo Wang et al.
The Journal of biological chemistry, 282(17), 12610-12618 (2007-03-06)
The core human mitochondrial transcription machinery comprises a single subunit bacteriophage-related RNA polymerase, POLRMT, the high mobility group box DNA-binding protein h-mtTFA/TFAM, and two transcriptional co-activator proteins, h-mtTFB1 and h-mtTFB2 that also have rRNA methyltransferase activity. Recapitulation of specific initiation
Valérie Serre et al.
Biochimica et biophysica acta, 1832(8), 1304-1312 (2013-04-23)
Multiple respiratory chain deficiencies represent a common cause of mitochondrial diseases and are associated with a wide range of clinical symptoms. We report a subject, born to consanguineous parents, with growth retardation and neurological deterioration. Multiple respiratory chain deficiency was
L Marty et al.
Genomics, 41(3), 453-457 (1997-05-01)
Mitochondrial activity requires the expression of nuclear genes, whose products are part of multiproteic complexes leading to ATP production and delivery. We recently characterized a growth-activated mRNA encoding the human mitochondrial ribosomal MRPL12 protein, which is thought to act as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service