Skip to Content
Merck
All Photos(4)

Key Documents

SAB1400284

Sigma-Aldrich

Monoclonal Anti-TTN antibody produced in mouse

clone 7D3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CMD1G, Anti-CMH9, Anti-CMPD4, Anti-DKFZp451N061, Anti-FLJ32040, Anti-LGMD2J, Anti-TMD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7D3, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TTN(7273)

General description

This gene encodes a large abundant protein of striated muscle. The product of this gene is divided into two regions, a N-terminal I-band and a C-terminal A-band. The I-band, which is the elastic part of the molecule, contains two regions of tandem immunoglobulin domains on either side of a PEVK region that is rich in proline, glutamate, valine and lysine. The A-band, which is thought to act as a protein-ruler, contains a mixture of immunoglobulin and fibronectin repeats, and possesses kinase activity. A N-terminal Z-disc region and a C-terminal M-line region bind to the Z-line and M-line of the sarcomere respectively so that a single titin molecule spans half the length of a sarcomere. Titin also contains binding sites for muscle associated proteins so it serves as an adhesion template for the assembly of contractile machinery in muscle cells. It has also been identified as a structural protein for chromosomes. Considerable variability exists in the I-band, the M-line and the Z-disc regions of titin. Variability in the I-band region contributes to the differences in elasticity of different titin isoforms and, therefore, to the differences in elasticity of different muscle types. Of the many titin variants identified, five for which complete transcript information is available are described. Mutations in this gene are associated with familial hypertrophic cardiomyopathy 9 and autoantibodies to titin are produced in patients with the autoimmune disease scleroderma. (provided by RefSeq)

Immunogen

TTN (AAH58824, 1 a.a. ~ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MTTQAPTFTQPLQSVVVLEGSTATFEAHISGFPVPEVSWFRDGQVISTSTLPGVQISFSDGRAKLTIPAVTKANSGRYSLKATNGSGQATSTAELLVKAETAPPNFVQRL

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aurélien Perrin et al.
Annals of clinical and translational neurology, 8(9), 1906-1912 (2021-07-28)
The aim of this study was to analyze patients from two distinct families with a novel distal titinopathy phenotype associated with exactly the same CNV in the TTN gene. We used an integrated strategy combining deep phenotyping and complete molecular
Meng Yu et al.
Annals of clinical and translational neurology, 6(7), 1311-1318 (2019-07-30)
We present clinical features, muscle imaging findings, and genetic characteristics of five unrelated Chinese patients with congenital titinopathy, emphasizing the diagnostic role of muscle MRI. Five patients who recessive titinopathies were recruited. All patients received muscle biopsies. Mutations were detected
Aurélien Perrin et al.
Neuromuscular disorders : NMD, 30(11), 877-887 (2020-11-01)
Next generation sequencing (NGS) has allowed the titin gene (TTN) to be identified as a major contributor to neuromuscular disorders, with high clinical heterogeneity. The mechanisms underlying the phenotypic variability and the dominant or recessive pattern of inheritance are unclear.
Jianing Chen et al.
Frontiers in oncology, 12, 877878-877878 (2022-07-26)
Lung adenocarcinoma (LUAD) is the most common type of lung cancer, accounting for around 40%. Despite achievements in the treatment approach, the prognosis is still dismal, with overall survival of fewer than five years. Thus, novel prognostic biomarkers are needed
Ozge Ceyhan-Birsoy et al.
Neurology, 81(14), 1205-1214 (2013-08-27)
To identify causative genes for centronuclear myopathies (CNM), a heterogeneous group of rare inherited muscle disorders that often present in infancy or early life with weakness and hypotonia, using next-generation sequencing of whole exomes and genomes. Whole-exome or -genome sequencing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service