Skip to Content
Merck
All Photos(1)

Documents

SAB1300678

Sigma-Aldrich

Anti-KLF4 antibody produced in rabbit

saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution

Synonym(s):

Anti-EZF, Anti-GKLF, Anti-Kruppel-like factor 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

saturated ammonium sulfate (SAS) precipitated

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KLF4(9314)

General description

Krüppel-like factor 4 (KLF4) is a transcription factor. It can act both as a transcriptional activator or repressor. The gene encoding this protein is localized on human chromosome 9q31.2.

Immunogen

KLF4 (NP_004226.3, )
This antibody is generated from rabbits immunized with a his tag recombinant protein of human KLF4.

Biochem/physiol Actions

Krüppel-like factor 4 (KLF4) is involved in cell growth, proliferation and differentiation in the colon. It is down-regulated in both mouse and human colonic adenomas. The protein has been implicated as a tumor suppressor in the gut, whereas in breast cancer, KLF4 is an oncogene. It is also involved in reprogramming differentiated cells into pluripotent stem cells.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dynamic down-regulation of Kruppel-like factor 4 in colorectal adenoma-carcinoma sequence.
Xu J
Journal of Cancer Research and Clinical Oncology (2008)
Kruppel-like factor 4 (KLF4): What we currently know.
Ghaleb AM and Yang VW
Gene (2017)
Advances in Reprogramming Somatic Cells to Induced Pluripotent Stem Cells
Minal Patel
Stem Cell Reviews (2010)
Kruppel-like factor 4 is acetylated by p300 and regulates gene transcription via modulation of histone acetylation.
Evans PM
The Journal of Biological Chemistry (2007)
KLF4 is a Novel Candidate Tumor Suppressor Gene in Pancreatic Ductal Carcinoma
Francesca Zammarchi
The American Journal of Pathology (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service