Skip to Content
Merck
All Photos(3)

Key Documents

SAB1300113

Sigma-Aldrich

Anti-TGFBR1 (center) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

TGF-β receptor type I, TGFR-1, TGF-β type I receptor, Serine/threonine-protein kinase receptor R4, SKR4, Activin receptor-like kinase 5, ALK-5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TGFBR1(7046)

General description

TGFBR1 (transforming growth factor β receptor 1) gene is mapped to human chromosome 9q22.33. TGFBR1 is known to localize in the focal adhesions of the cell surface.
The protein encoded by this gene forms a heteromeric complex with type II TGF-β receptors when bound to TGF-beta, transducing the TGF-beta signal from the cell surface to the cytoplasm. The encoded protein is a serine/threonine protein kinase. Mutations in this gene have been associated with Loeys-Dietz aortic aneurysm syndrome (LDAS).

Immunogen

TGFBR1 (NP_004603, 138-173)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the central region of human TGFBR1.

Application

Anti-TGFBR1 (center) antibody produced in rabbit has been used in western blot.

Biochem/physiol Actions

TGFBR1 (transforming growth factor β receptor 1) stimulates signaling pathway that mediates cell growth and division. The complex formed by TGFBR2 with TGFBR1 results in the phosphorylation and activation of TGFBR1. TGFBR1 is responsible for the activation of canonical Smad (mothers against decapentaplegic homolog 3) proteins and transcriptional factors Snail (zinc finger protein) and Twist (twist family bHLH transcription factor 1). The activation of a number of non-canonical effectors such as RhoA (ras homolog family member A), TAK1 (transforming growth factor-β-activated kinase 1) and Akt (RAC-α serine/threonine-protein kinase) is also associated with TGFBR1. The arrangement of the TGFBR1 receptor along with TGFBR2 at the focal adhesions mediates the multimerization and stimulation of TGFβ ligand. Downregulation of TGFBR1 is observed in breast cancer. TGFBR1 is also known to be associated with colorectal cancer.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ibrahim Y Hachim et al.
Human pathology, 57, 140-151 (2016-10-25)
The transforming growth factor-β (TGFβ) plays a dual role in breast cancer, acting as a tumor suppressor in early carcinomas while promoting tumor metastasis in more advanced breast carcinoma. As a result, the prognostic role of TGFβ and its signaling
Rosa M Xicola et al.
Carcinogenesis, 37(8), 751-758 (2016-05-29)
The purpose of this study was to identify novel colorectal cancer (CRC)-causing alleles in unexplained familial CRC cases. In order to do so, coding regions in five candidate genes (MGMT, AXIN2, CTNNB1, TGFBR1 and TGFBR2) were sequenced in 11 unrelated
Li Yang et al.
PloS one, 11(5), e0155270-e0155270 (2016-05-14)
Endometrial cancer (EC) is a complex disease involving multiple gene-gene and gene-environment interactions. TGF-β signaling plays pivotal roles in EC development. This study aimed to investigate whether the genetic polymorphisms of TGF-β signaling related genes TGFB1, TGFBR1, SNAI1 and TWIST1
Joanna P Rys et al.
eLife, 4, e09300-e09300 (2015-12-15)
Cell surface receptors are central to the cell's ability to generate coordinated responses to the multitude of biochemical and physical cues in the microenvironment. However, the mechanisms by which receptors enable this concerted cellular response remain unclear. To investigate the
Sergiy Klid et al.
Stem cell research & therapy, 12(1), 608-608 (2021-12-22)
An environment of gestational diabetes mellitus (GDM) can modify the phenotype of stem cell populations differentially according to their placental localization, which can be useful to study the consequences for the fetus. We sought to explore the effect of intrauterine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service