Skip to Content
Merck
All Photos(1)

Key Documents

S8446

Sigma-Aldrich

SIRT1 Peptide

≥90% (SDS-PAGE), human recombinant, expressed in E. coli, N-terminal histidine tagged

Synonym(s):

SIR2α, SIR2L1, Sirtuin1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

Product Name

SIRT1 human, recombinant, expressed in E. coli, N-terminal histidine tagged, ≥90% (SDS-PAGE), buffered aqueous glycerol solution

recombinant

expressed in E. coli

Quality Level

description

full-length amino acid sequence of original SIRT1 protein (accession number NP_036370)

Assay

≥90% (SDS-PAGE)

form

buffered aqueous glycerol solution

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... SIRT1(23411)

General description

SIRT1 (sirtuin 1) belongs to the silent information regulator (SIR) family and is a class of HDAC (histone deacetylase). It is expressed in various tissues including brain, liver, pancreas, adipose tissue, and skeletal muscle. SIRT1 is also known as the longevity gene.

Application

Human SIRT1 (sirtuin1) has been used in in vitro acetylation and deacetylation assays. It has also been used to study its effects on homeostasis of human nucleus pulposus cells.

Biochem/physiol Actions

SIRT1 (sirtuin 1) functions as a regulator of various metabolic pathways, and influences the pathophysiology of several metabolic diseases. It is a regulator of protein deacetylation, and is a candidate therapeutic target in non-alcoholic fatty liver disease (NAFLD), amyotrophic lateral sclerosis (ALS), kidney disease, and pulmonary disease. It also participates in tumorigenesis, and whether it functions as an oncogene or as a tumor suppressor depends upon the tumor type. In pancreatic ductal adenocarcinoma (PDAC) its elevated expression is linked with poor prognosis, and in non-small-cell lung cancer (NSCLC) it suppresses the expression of tumor suppressor p27. It is also thought to function as a suppressor of cardiovascular disorders, such as myocardial infarction, or neurodegenerative diseases, such as Alzheimer′s disease (AD) or Parkinson′s disorder (PD).
SIRT1 plays a pivotal role in the regulation of cellular differentiation, metabolism, cell cycle, apoptosis and regulation of p53.
Sirtuins are a family of NAD+ dependent deacetylases that remove an acetyl group from the e-amino group of lysine residues. The proteins within this family are named after the first protein discovered, from yeast, called Sir2 (Silent Information Regulator 2). The proteins are conserved from bacteria to higher eukaryotes. In humans, there are seven Sir2 family members (SIRT1 to SITR7). SIRT1 plays a pivotal role in the regulation of cellular differentiation, metabolism, cell cycle, apoptosis and regulation of p53. Several targets for SIRT1 were identified among them Lys382 of p53. Using RNA interference, additional targets were identified. It was demonstrated that reduced levels of human SIRT1 led to increased acetylation of Histone H4-Lys16, H4-Lys20, and Histone H3-Lys9 as well as histone H1-Lys26.

Physical form

Solution containing 50 mM Tris, pH 7.4, 100 mM NaCl, 1 mM DTT, protease inhibitors (Product code P8340) 1:200, and 10% glycerol (w/v).

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression of silent mating type information regulator 2 homolog 1 and its role in human intervertebral disc cell homeostasis
Arthritis Research & Therapy (2011)
Modulations of hMOF autoacetylation by SIRT1 regulate hMOF recruitment and activities on the chromatin
Lu Lu
Cell Research (2011)
Yeon-Hwa Lee et al.
Cancer letters, 431, 219-229 (2018-05-29)
SIRT1, an NAD+-dependent histone/protein deacetylase, has diverse physiological actions. Recent studies have demonstrated that SIRT1 is overexpressed in colorectal cancer, suggesting its oncogenic potential. However, the molecular mechanisms by which overexpressed SIRT1 induces the progression of colorectal cancer and its
Taek-Yeol Jung et al.
Experimental & molecular medicine, 52(7), 1075-1089 (2020-07-09)
Histidine triad nucleotide-binding protein 1 (HINT1), which belongs to the evolutionarily conserved HIT superfamily, has been shown to possess a tumor-suppressive function by binding to and inhibiting several oncogenic transcription factors, such as β-catenin and microphthalmia transcription factor (MITF), in
Chae Jin Lim et al.
Biomolecules & therapeutics, 25(5), 511-518 (2017-08-22)
Ultraviolet (UV) irradiation is a relevant environment factor to induce cellular senescence and photoaging. Both autophagy- and silent information regulator T1 (SIRT1)-dependent pathways are critical cellular processes of not only maintaining normal cellular functions, but also protecting cellular senescence in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service