Skip to Content
Merck
All Photos(1)

Key Documents

S6402

Sigma-Aldrich

Streptavidin−Cy3 from Streptomyces avidinii

buffered aqueous solution

Synonym(s):

Streptavidin Cy3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

conjugate

CY3 conjugate

form

buffered aqueous solution

extent of labeling

3-9 mol fluorochrome per mol protein

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100

shipped in

wet ice

storage temp.

2-8°C

General description

Streptavidin is a homotetramer protein. One molecule of streptavidin binds four molecules of biotin by a non-covalent interaction that is essentially irreversible. Streptavidin has a high affinity for biotin and is extensively used as a labeling tool and for studies involving binding proteins.

Application

Streptavidin−Cy3 from Streptomyces avidinii has been used:
  • in double immunofluorescence staining to detect biotinylated lectin
  • for insulin and C-peptide staining in immunofluorescence analysis
  • in double immunofluorescence to verify the presence of cells filled with biocytin
  • in conjunction with biotinylated antibodies in immunohistochemistry and immunocytochemistry
  • with the traditional rhodamine filter arrangements due to the spectral overlap

Features and Benefits

Spectral Characteristics of Cy3 Dye:
  • Absorbance Max 552 nm
  • Emission Max 565 nm
  • Molar Extinction Coefficient 1.3 x 10-5 M-1cm-1

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin, and 15 mM sodium azide

Legal Information

Cy3 is a trademark of Cytiva

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ji-Myung Bae et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 33(6), 1126-1140 (2018-02-07)
The Sp7/Osterix transcription factor is essential for bone development. Mutations of the Sp7 gene in humans are associated with craniofacial anomalies and osteogenesis imperfecta. However, the role of Sp7 in embryonic tooth development remains unknown. Here we identified the functional
Hiroyuki Nobusue et al.
Cell and tissue research, 332(3), 435-446 (2008-04-04)
We established a preadipocyte cell line from mature adipocytes obtained from subcutaneous fat tissue of green fluorescent protein (GFP) transgenic mice. The floating top layer, containing mature adipocytes, was isolated from subcutaneous fat tissue by collagenase digestion and filtration. Fluorescence-activated
Agnieszka E Gorska et al.
The American journal of pathology, 163(4), 1539-1549 (2003-09-26)
We have previously shown that expression of a dominant-negative type II transforming growth factor-beta receptor (DNIIR) in mammary epithelium under control of the MMTV promoter/enhancer causes alveolar hyperplasia and differentiation in virgin mice. Here we show that MMTV-DNIIR female mice
Niklas Schwarz et al.
eLife, 8 (2019-09-10)
Most of our knowledge on human CNS circuitry and related disorders originates from model organisms. How well such data translate to the human CNS remains largely to be determined. Human brain slice cultures derived from neurosurgical resections may offer novel
Kathleen R Makielski et al.
Virology, 495, 52-62 (2016-05-15)
Epstein-Barr virus and human papillomaviruses are human tumor viruses that infect and replicate in upper aerodigestive tract epithelia and cause head and neck cancers. The productive phases of both viruses are tied to stratified epithelia highlighting the possibility that these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service