Skip to Content
Merck
All Photos(3)

Documents

S5375

Sigma-Aldrich

Shikimic acid

≥99%, suitable for ligand binding assays

Synonym(s):

(3R,4S,5R)-(−)-3,4,5-Trihydroxy-1-cyclohexenecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10O5
CAS Number:
Molecular Weight:
174.15
Beilstein:
2210055
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

product name

Shikimic acid, ≥99%

Assay

≥99%

form

powder

technique(s)

ligand binding assay: suitable

color

white to off-white

mp

185-187 °C (lit.)

SMILES string

O[C@@H]1CC(=C[C@@H](O)[C@H]1O)C(O)=O

InChI

1S/C7H10O5/c8-4-1-3(7(11)12)2-5(9)6(4)10/h1,4-6,8-10H,2H2,(H,11,12)/t4-,5-,6-/m1/s1

InChI key

JXOHGGNKMLTUBP-HSUXUTPPSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Shikimic acid has been used as a standard for the quantification of shikimate in apical parts of roots. It has also been used as a substrate in shikimate kinase assay.

Biochem/physiol Actions

Shikimic acid is observed to be carcinogenic in rat models, when administered. It serves as a precursor for the biosynthesis of aromatic amino acids, alkaloids and other aromatic metabolites in microorganisms. Shikimic acid is known to inhibit adenosine diphosphate (ADP) induced platelet aggregation and blood coagulation in rabbits.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient elimination of nonstoichiometric enzyme inhibitors from HTS hit lists.
Habig M
Journal of Biomolecular Screening, 14(6), 679-689 (2009)
Kai Chen et al.
Bioresource technology, 119, 141-147 (2012-06-26)
Shikimic acid (SA) is an important metabolic intermediate with diverse commercial applications. In this work, antisense RNA interference and gene deletion were carried out to inactivate the aroK gene in an SA-producing Escherichia coli strain, DHPYA-T7. In this strain, the
Varun Rawat et al.
Organic & biomolecular chemistry, 10(20), 3988-3990 (2012-04-24)
A new enantioselective synthesis of the anti-influenza agent (-)-oseltamivir free base (7.1% overall yield; 98% ee) and (-)-methyl 3-epi-shikimate (16% overall yield; 98% ee) has been described from readily available raw materials. Sharpless asymmetric epoxidation and diastereoselective Barbier allylation of
Glyphosate inhibition of ferric reductase activity in iron deficient sunflower roots.
Ozturk L
The New phytologist, 177(4), 899-906 (2008)
Amalia M Estévez et al.
Mini reviews in medicinal chemistry, 12(14), 1443-1454 (2012-07-26)
Shikimic acid, a natural compound is a key intermediate in the biosynthesis of amino acids. Consequently, this derivative is widely present in many plants and has interesting biological properties. But besides the pharmacological relevance of shikimic acid itself, it is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service