Skip to Content
Merck
All Photos(3)

Key Documents

S4045

Sigma-Aldrich

Monoclonal Anti-Splicing Factor SC-35 antibody produced in mouse

clone SC-35, ascites fluid

Synonym(s):

Anti-SC-35, Anti-SC35

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

SC-35, monoclonal

mol wt

antigen 35 kDa (a doublet)

contains

15 mM sodium azide

species reactivity

frog, Drosophila, newt, rat, human

technique(s)

electron microscopy: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
indirect immunofluorescence: 1:2,000 using cultured human fibroblasts

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SFRS2(6427)
rat ... Sfrs2(494445)

General description

Monoclonal Anti-Splicing Factor SC-35 (mouse IgG1 isotype) is derived from the SC-35 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized RBF-DNJ mouse. Splicing component of 35 kDa (SC-35) is also termed as PR264. SC-35 displays a speckled distribution in the nucleus that co-localizes with small nuclear ribonucleoproteins (snRNPs), but unlike snRNPs, SC-35 does not give diffuse nuclear labeling.
Nuclear pre-mRNA splicing takes place in a multi-component structure termed a spliceosome. Major subunits of spliceosomes are U1, U2, U4/U6 and U5 small nuclear ribonucleoproteins (snRNP′s). In addition to the snRNP′s, a number of protein factors have been identified which are required for spliceosome assembly and splicing. For example, the protein factors U2AF, SF2 and SF3 are required for the binding of the U2 snRNP to the intron branch-point and for assembly of the pre-splicing complex. Two other non-snRNP splicing factors, SF2/ASF and SC-35 (splicing component of 35 kD, also termed PR264), are both required for the first step of splicing and spliceosome assembly. SF2/ASF and SC-35 are also involved in 5N splice site selection of alternatively spliced pre-mRNA′s.
The essential non-snRNP splicing factor SC-35 displays a speckled distribution in the nucleus that co-localizes with snRNPs, but unlike snRNPs, SC-35 does not give diffuse nuclear labelling. In the nucleus, snRNPs are concentrated in coiled bodies and in the speckled regions, whereas SC-35 is found in speckles but not in coiled bodies.

Specificity

Recognizes a phospho-epitope on the non-snRNP (small nuclear ribonucleoprotein particles) factor SC-35. The antibody reacts with the splicing factor SC-35 and with the SC-35-related non-snRNP factor SF2/ASF. The antibody labels SC-35 as a speckled pattern in the nucleoplasm, excluding the nucleoli. Other uses include inhibition and depletion of splicing activity in nuclear extracts and immunoaffinity purification.

Immunogen

partially purified mammalian splicesome.

Application

Monoclonal Anti-Splicing Factor SC-35 antibody produced in mouse has been used in:
  • fluorescence image analysis
  • immunohistochemistry
  • two or three color immunofluorescence staining
  • enzyme-linked immunosorbent assay (ELISA)
  • immunohistology
  • immunoelectronmicroscopy
  • immunoaffinity purification
  • immunoprecipitation

Monoclonal Anti-Splicing Factor SC-35 may be used for the localization of SC-35 using ELISA, immunohistology and immunoelectronmicroscopy, for inhibition and depletion of splicing activity in nuclear extracts, and for immunoaffinity purification and immunoprecipitation.
Indirect immunofluorescence: a dilution of at least 1:2,000 was determined by staining cultured human fibroblasts.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization and cloning of the human splicing factor 9G8: a novel 35 kDa factor of the serine/arginine protein family.
Cavaloc Y, et al.
The Embo Journal, 13(11), 2639-2649 (1994)
X D Fu et al.
Nature, 343(6257), 437-441 (1990-02-01)
A monoclonal antibody raised against mammalian spliceosomes specifically recognizes a non-snRNP factor required for spliceosome assembly. This splicing factor is highly concentrated in discrete regions within the nucleus, in a pattern that is a distinct subset of that seen with
Silvia Vivarelli et al.
PloS one, 8(4), e61980-e61980 (2013-04-25)
Paraquat (PQ) is a neurotoxic herbicide that induces superoxide formation. Although it is known that its toxic properties are linked to ROS production, the cellular response to PQ is still poorly understood. We reported previously that treatment with PQ induced
Yasmine Even et al.
PloS one, 11(2), e0149184-e0149184 (2016-02-18)
The perinucleolar compartment (PNC) is a subnuclear stucture forming predominantly in cancer cells; its prevalence positively correlates with metastatic capacity. Although several RNA-binding proteins have been characterized in PNC, the molecular function of this compartment remains unclear. Here we demonstrate
X D Fu et al.
Proceedings of the National Academy of Sciences of the United States of America, 89(23), 11224-11228 (1992-12-01)
The human pre-mRNA splicing factors SF2 and SC35 have similar electrophoretic mobilities, and both of them contain an N-terminal ribonucleoprotein (RNP)-type RNA-recognition motif and a C-terminal arginine/serine-rich domain. However, the two proteins are encoded by different genes and display only

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service