Skip to Content
Merck
All Photos(1)

Key Documents

S3572

Sigma-Aldrich

Stevioside hydrate

from Stevia sp., ≥98% (HPLC), powder, p-aminohippurate transepithelial transport inhibitor

Synonym(s):

(4α-13-[(2-O-β-D-Glucopyranosyl-β-D-glucopyranosyl)oxy]kaur-16-en-18-oic acid β-D-glucopyranosyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H60O18 · xH2O
CAS Number:
Molecular Weight:
804.87 (anhydrous basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Stevioside hydrate, ≥98% (HPLC)

biological source

Stevia sp.

Assay

≥98% (HPLC)

form

powder

color

white

solubility

DMSO: 25 mg/mL, clear

storage temp.

2-8°C

SMILES string

O.C[C@@]12CCC[C@](C)(C1CC[C@]34CC(=C)[C@](CCC23)(C4)O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O)C(=O)O[C@@H]7O[C@H](CO)[C@@H](O)[C@H](O)[C@H]7O

InChI

1S/C38H60O18.H2O/c1-16-11-37-9-5-20-35(2,7-4-8-36(20,3)34(50)55-32-29(49)26(46)23(43)18(13-40)52-32)21(37)6-10-38(16,15-37)56-33-30(27(47)24(44)19(14-41)53-33)54-31-28(48)25(45)22(42)17(12-39)51-31;/h17-33,39-49H,1,4-15H2,2-3H3;1H2/t17-,18-,19-,20+,21+,22-,23-,24-,25+,26+,27+,28-,29-,30-,31+,32+,33+,35-,36-,37-,38+;/m1./s1

InChI key

FVECWZKNZGXJJO-QMYNVHDOSA-N

Application

Stevioside hydrate has been used as a standard in high performance liquid chromatography (HPLC) for quantification of steviol glycosides.
Stevioside-induced ROS-mediated apoptosis was studied in human breast cancer cells.

Biochem/physiol Actions

Stevioside hydrate is a diterpene glycoside present in Stevia sp.
Stevioside is a noncaloric natural sweetener, 300 times more potent than sucrose. It inhibits transepithelial transport of p-aminohippurate (PAH) by interfering with the organic anion transport system. At 0.5-1 mM, it showed no interaction with any organic anion transporters (OAT).
Stevioside reportedly has genotoxic effects in cultured mammalian cells.

Preparation Note

Stevioside solutions should be prepared in DMSO. Highly pure stevioside has very poor water solubility.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tadamasa Terai et al.
Chemical & pharmaceutical bulletin, 50(7), 1007-1010 (2002-07-20)
Stevioside is natural non-caloric sweetner isolated from Stevia rebaudiana BERTONI, which has been used as a non-caloric sugar substitute in Japan. Pezzuto et al. demonstrated that steviol shows a dose-dependent positive response in forward mutation assay using Salmonella typhimurium TM677
J M Pezzuto et al.
Proceedings of the National Academy of Sciences of the United States of America, 82(8), 2478-2482 (1985-04-01)
Stevioside, a constituent of Stevia rebaudiana, is commonly used as a noncaloric sugar substitute in Japan. Consistent with reports in the literature, we have found that stevioside is not mutagenic as judged by utilization of Salmonella typhimurium strain TM677, either
Evaluation of the euglycemic effect of oral administration of S. rebaudiana B. cultivated in Mexico in normoglycemic and induced-diabetic rats
Irma AG, et al.
International journal of environment, agriculture and biotechnology, 2(1) null
Determination of rebaudioside A and Stevioside in leaves of S. rebaudiana Bertoni grown in Mexico by a validated HPLC method
Aranda-Gonzalez I, et al.
American Journal of Analytical Chemistry, 6(11), 878-878 (2015)
Stevioside and related sweet diterpenoid glycosides.
J R Hanson et al.
Natural product reports, 10(3), 301-309 (1993-06-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service