Skip to Content
Merck
All Photos(1)

Key Documents

S1069

Sigma-Aldrich

(S)-SNAP-5114

≥98% (HPLC), solid

Synonym(s):

(S)-1-[2-[Tris(4-methoxyphenyl)methoxy]ethyl]-3-piperidinecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H35NO6
CAS Number:
Molecular Weight:
505.60
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

optical activity

[α]/D -8 to -13°, c = 1 in chloroform-d

color

white

solubility

DMSO: >5 mg/mL
H2O: insoluble

storage temp.

−20°C

SMILES string

COc1ccc(cc1)C(OCCN2CCC[C@@H](C2)C(O)=O)(c3ccc(OC)cc3)c4ccc(OC)cc4

InChI

1S/C30H35NO6/c1-34-26-12-6-23(7-13-26)30(24-8-14-27(35-2)15-9-24,25-10-16-28(36-3)17-11-25)37-20-19-31-18-4-5-22(21-31)29(32)33/h6-17,22H,4-5,18-21H2,1-3H3,(H,32,33)/t22-/m0/s1

InChI key

VDLDUZLDZBVOAS-QFIPXVFZSA-N

Biochem/physiol Actions

GABA transport inhibitor, showing selectivity for GAT-3 and GAT-2.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the GABA Transporters page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Ek Lie et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 38(1), 166-173 (2017-11-18)
Brain ischemia triggers excitotoxicity and cell death, yet no neuroprotective drugs have made it to the clinic. While enhancing GABAergic signaling to counterbalance excitotoxicity has shown promise in animal models, clinical studies have failed. Blockade of GABA transporters (GATs) offers
Jonas M Fuks et al.
PLoS pathogens, 8(12), e1003051-e1003051 (2012-12-14)
During acute infection in human and animal hosts, the obligate intracellular protozoan Toxoplasma gondii infects a variety of cell types, including leukocytes. Poised to respond to invading pathogens, dendritic cells (DC) may also be exploited by T. gondii for spread
N O Dalby
Neuropharmacology, 39(12), 2399-2407 (2000-09-07)
The present study examines the effect of tiagabine (a selective inhibitor of GABA transporter 1, GAT-1), SNAP-5114 (a semi-selective inhibitor of rat GAT-3/mouse GAT4) and NNC 05-2045 (a non-selective GABA uptake inhibitor) in modulating GABA levels in the hippocampus and
Adam C Lu et al.
eLife, 9 (2020-09-10)
Absence seizures result from 3 to 5 Hz generalized thalamocortical oscillations that depend on highly regulated inhibitory neurotransmission in the thalamus. Efficient reuptake of the inhibitory neurotransmitter GABA is essential, and reuptake failure worsens human seizures. Here, we show that
L A Borden
Neurochemistry international, 29(4), 335-356 (1996-10-01)
gamma-Aminobutyric acid (GABA) is the major inhibitory neurotransmitter in the mammalian brain. GABA is cleared from the synaptic cleft by specific, high-affinity, sodium- and chloride-dependent transporters, which are thought to be located on presynaptic terminals and surrounding glial cells. While

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service