Skip to Content
Merck

RPROB

Reversible Protein Detection Kit for Membranes and Polyacrylamide Gels

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

NACRES:
NA.32
UNSPSC Code:
12352200
Technique(s):
protein staining: suitable
Usage:
sufficient for 20 applications staining (small 8 X 10 cm blots or PAGE gels)
Storage temp.:
2-8°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

usage

sufficient for 20 applications staining (small 8 X 10 cm blots or PAGE gels)

Quality Level

technique(s)

protein staining: suitable

storage temp.

2-8°C

Application

R-PROB is a unique protein detection system designed for staining of proteins on nylon, nitrocellulose, and PVDF membranes or PAGE gels with the detection sensitivity similar to that of Coomassie stains. Most other stains produce high backgrounds on nylon membranes due to strong charge interactions with the membrane. The mixing of equal amounts of Solution A and B activates the stain, which produces lavender stained protein bands. The staining is easily reversible with an EDTA solution so that the blot can be reused for Western blotting or for amino acid sequence analysis (PVDF). R-PROB does not contain the toxic chemicals cacodylic acid or potassium ferrocyanide.

Other Notes

Compatible with nylon, nitrocellulose, and PVDF membranes as well as polyacrylamide gels.

Kit Components Only

Product No.
Description

  • Solution A 500 mL

  • Solution B 500 mL

pictograms

Corrosion

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sensitivity and mass accuracy for proteins analyzed directly from polyacrylamide gels: Implications for proteome mapping
Rachel R.
Electrophoresis (1997)

Related Content

Global Trade Item Number

SKUGTIN
RPROB-1KT04061835573967

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service