Skip to Content
Merck

RDD024

Tricine

anhydrous, free-flowing, Redi-Dri, ≥99%

Synonym(s):

N-[Tris(hydroxymethyl)methyl]glycine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(HOCH2)3CNHCH2CO2H
CAS Number:
Molecular Weight:
179.17
UNSPSC Code:
12161700
NACRES:
NA.25
PubChem Substance ID:
EC Number:
227-193-6
Beilstein/REAXYS Number:
1937804
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

anhydrous

Quality Segment

product line

Redi-Dri

assay

≥99%

form

powder

quality

free-flowing

impurities

≤0.2% water (Karl Fischer)

useful pH range

7.4-8.8

pKa (25 °C)

8.1

solubility

water: 20 % (w/w), clear, colorless

cation traces

heavy metals (as Pb): ≤5 ppm

absorption

≤0.02 at 290 nm at 20 % (w/w)

SMILES string

OCC(CO)(CO)NCC(O)=O

InChI

1S/C6H13NO5/c8-2-6(3-9,4-10)7-1-5(11)12/h7-10H,1-4H2,(H,11,12)

InChI key

SEQKRHFRPICQDD-UHFFFAOYSA-N

Application

Buffer component for separation of low molecular weight peptides.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC


Still not finding the right product?

Explore all of our products under Tricine


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library