Skip to Content
Merck
All Photos(1)

Key Documents

RAB0743

Sigma-Aldrich

Human HAVCR1 / Hepatitis A Virus Cellular Receptor 1 ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 2 pg/mL
standard curve range: 12.29-3000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... HAVCR1(26762)

General description

The antibody pair provided in this kit recognizes Human Hepatitis A Virus Cellular Receptor 1.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aya Imafuku et al.
Stem cells translational medicine, 8(12), 1330-1341 (2019-11-02)
Renal fibrosis is one of the largest global health care problems, and microvascular (MV) injury is important in the development of progressive fibrosis. Although conventional cell therapy suppresses kidney injury via the role of vasoprotective cytokines, the effects are limited
Yu-Hsing Chang et al.
Therapeutic advances in chronic disease, 11, 2040622320964161-2040622320964161 (2020-11-03)
Elevated plasma C-terminal fibroblast growth factor-23 (cFGF-23) levels are associated with higher mortality in patients with chronic kidney disease (CKD) and acute kidney injury (AKI). Our study explored the outcome forecasting accuracy of cFGF-23 in critically ill patients with CKD
Patric Schyman et al.
Frontiers in pharmacology, 9, 1272-1272 (2018-11-22)
Ingestion or exposure to chemicals poses a serious health risk. Early detection of cellular changes induced by such events is vital to identify appropriate countermeasures to prevent organ damage. We hypothesize that chemically induced organ injuries are uniquely associated with
Suzana Bojic et al.
Disease markers, 2018, 5064684-5064684 (2018-06-05)
The role of matrix metalloproteinase-9 (MMP-9) and tissue inhibitor of matrix metalloproteinase-1 (TIMP-1) in sepsis after major abdominal surgery and sepsis-associated organ dysfunction is unexplored. Fifty-three patients with sepsis after major abdominal surgery were compared to 50 operated and 50
Kevin J Downes et al.
The Journal of antimicrobial chemotherapy, 72(1), 254-260 (2016-09-03)
Tobramycin is frequently used for treatment of bronchopneumonia in patients with cystic fibrosis (CF). Variability in tobramycin clearance (CL) is high in this population with few reliable approaches to guide dosing. We sought to evaluate the pharmacokinetics of once-daily intravenous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service