Skip to Content
Merck
All Photos(1)

Key Documents

RAB0720

Sigma-Aldrich

Human PAPPA / Pappalysin-1 ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.6 ng/mL
standard curve range: 0.686-500 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PAPPA(5069)

General description

The PAPPA ELISA kit provides for the Quantitative measurement of Pappalysin-1 in Cell Culture Supernatants, Plasma and Serum.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erik Nilsson et al.
European journal of clinical investigation, 48(8), e12959-e12959 (2018-05-29)
Elevated pregnancy-associated plasma protein A (PAPP-A) levels are associated with increased risk of death in ischaemic heart disease as well as in haemodialysis patients. Previous research indicates that the prognostic value of PAPP-A may be stronger in patients with concomitant
Dursun Ali Sirin et al.
Odontology, 109(1), 250-258 (2020-07-01)
To examine the use of IL-6 and PAPP-A along with the new imaging for scoring methods to determine the severity of apical periodontitis (AP). The scoring of disease severity in 121 patients with AP, using dental radiographs, was based on
Chen Chen et al.
Annals of translational medicine, 8(9), 579-579 (2020-06-23)
The aim of this study was to evaluate serum biomarkers of systemic lupus erythematous (SLE) patients, with and without neuropsychiatric (NP) manifestation by high-resolution proteomic analysis. SLE patients with NP (NPSLE, n=9), SLE patients without NP (non-NPSLE, n=9) and healthy
Diogo Torres et al.
PloS one, 14(11), e0224564-e0224564 (2019-11-22)
Inhibition of pregnancy-associated plasma protein-A (PAPP-A), an upstream activator of the insulin-like growth factor (IGF) pathway, is known to augment sensitivity to platinum-based chemotherapy. This study further tests the efficacy of PAPP-A inhibition with a monoclonal antibody inhibitor (mAb-PA) in
Sonu Kashyap et al.
JCI insight, 5(4) (2020-01-29)
Autosomal dominant polycystic kidney disease (ADPKD) is the most common genetic cause of end-stage renal disease (ESRD). The treatment options for ADPKD are limited. We observed an upregulation in several IGF-1 pathway genes in the kidney of Pkd1RC/RC mice, a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service