The Human EPO ELISA (RAB0654) has not been tested for cross-reactivity to Darbepoetin and has not been determined if the kit can be used for this purpose.
RAB0654
Human EPO / Erythropoietin ELISA Kit
for serum, plasma, cell culture supernatants and urine
Synonym(s):
Erythropoietin
About This Item
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
input
sample type cell culture supernatant(s)
sample type plasma
sample type serum
sample type urine
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.15 mIU/mL
standard curve range: 0.137-100 mIU/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... EPO(2056)
Related Categories
General description
Application
Biochem/physiol Actions
Other Notes
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Can this kit be used to determine quantitatively Darbepoeitin in Human Plasma ? If Yes what wpould becsensitivity.
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service