Skip to Content
Merck
All Photos(2)

Key Documents

RAB0292

Sigma-Aldrich

Mouse IL-28A/B ELISA

for serum, plasma and cell culture supernatant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

Quality Level

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)

UniProt accession no.

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 2 pg/mL
standard curve range: 2.46-600 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... , Ifnl2(282617)

General description

The Mouse IL-28A/B 5ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of Mouse IL-28A/B (IFN-lambda 2/3) in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Mouse IL-28B

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABELADCELISA 1X Assay/Sample Diluent Buffer C (Item L)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin-Sung Chung et al.
The Journal of investigative dermatology, 134(11), 2784-2794 (2014-06-18)
A major barrier to successful cancer immunotherapy is the tumor's ability to induce T-cell tolerance by exploiting host regulatory mechanisms. Having discovered the DC-HIL receptor, which inhibits T-cell responses by binding to syndecan-4 on effector T cells, we posited the
D M Bailey et al.
Acta physiologica (Oxford, England), 212(2), 175-187 (2014-05-09)
The aim of this study was to examine if erythropoietin (EPO) has the potential to act as a biological antioxidant and determine the underlying mechanisms. The rate at which its recombinant form (rHuEPO) reacts with hydroxyl (HO˙), 2,2-diphenyl-1-picrylhydrazyl (DPPH˙) and
Ill-Hwa Kim et al.
Theriogenology, 82(3), 427-432 (2014-06-17)
The objective of this study was to compare the concentrations of inflammatory cytokines in uterine flush and serum from healthy postpartum dairy cows and cows with clinical or subclinical endometritis. Clinical endometritis was diagnosed by observation of vaginal discharges (>50%
Ryan W Walker et al.
Obesity (Silver Spring, Md.), 22(6), 1512-1519 (2014-03-13)
Obesity in childhood is associated with an inflammatory state in adipose tissue and liver, which elevates risk for diabetes and liver disease. No prior study has examined associations between pathologies occurring in adipose tissue and liver to identify elements of
Jere Lindén et al.
International journal of molecular sciences, 15(8), 13938-13966 (2014-08-15)
One of the hallmarks of the acute toxicity of 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD) is a drastically reduced feed intake by an unknown mechanism. To further elucidate this wasting syndrome, we followed the effects of a single large dose (100 μg/kg) of TCDD

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service