Skip to Content
Merck
All Photos(2)

Documents

RAB0275

Sigma-Aldrich

Mouse IL-1 β ELISA Kit

for cell and tissue lysates

Synonym(s):

Cytokine detection kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type cell lysate
sample type tissue lysate

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 5 pg/mL
standard curve range: 2.74-2000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Il1b(16176)

General description

The Mouse IL-1 β ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse IL-1 β cell lysate and tissue lysate.

Immunogen

Recombinant Mouse IL-1 β

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Mouse IL-1 β ELISA Kit has been used to measure the concentration of interleukin 1 (IL)1-β in serum and bronchoalveolar lavage fluid (BALF) sample.

Biochem/physiol Actions

Interleukin 1 β (IL-1β) stimulates the production of endogenous 92 kDa gelatinase (matrix metalloproteinases 9 (MMP-9)) and tissue inhibitors of MMP (TIMP -1). It is a proinflammatory cytokine that induces various acute and chronic inflammation. Therefore, IL-1β can be considered as a potential therapeutic target for disease associated with fibrosis and tissue remodeling. IL-1β brings neutrophil and macrophage to the site of infection. Over expression of IL-1β in synovial fluid contributes to the pathogenesis of synovitis and degenerative changes of the cartilaginous tissue and bone of the temporomandibular joint. Mutations in IL-1β gene leads to renal manifestations and renal sequelae in Henoch-Schonlein purpura (HSP).

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABDIL6ELISA 5X Sample Diluent Buffer (Item D2)SDS

  • RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS

  • RABLYSIS12X Cell Lysis Buffer (Item J)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inactivation of the tyrosine phosphatase SHP-2 drives vascular dysfunction in Sepsis
Heun Y, et al.
EBioMedicine (2019)
Protective effects of fucoidan against hyperoxic lung injury via the ERK signaling pathway
Nie M, et al.
Molecular Medicine Reports, 17(1), 1813-1818 (2018)
Interleukin 1 beta (IL1?) rs16944 genetic variant as a genetic marker of severe renal manifestations and renal sequelae in Henoch-Schonlein purpura.
Lopez-Mejias R
Clinical and Experimental Rheumatology, 34(3 Suppl 97), S84-S88 (2016)
Proinflammatory cytokines detectable in synovial fluids from patients with temporomandibular disorders
Takahashi T, et al.
Oral Surgery, Oral Medicine, and Oral Pathology, 85(2), 135-141 (1998)
Strain- and host species-specific inflammasome activation, IL-1? release, and cell death in macrophages infected with uropathogenic Escherichia coli.
Schaale K
Mucosal Immunology, 9(1), 124-136 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service