RAB0216
Human HSP70 ELISA Kit
for serum, plasma, cell culture supernatant and urine
Synonym(s):
HSP70-1, Heat shock 70 kDa protein 1A
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type plasma
sample type urine
sample type cell culture supernatant(s)
sample type serum
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 2 ng/mL
standard curve range: 2.469-600 ng/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... HSPA4(3308)
General description
The Human HSP70 (heat shock protein 70) ELISA (enzyme-linked immunosorbent assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human HSP70 in serum, plasma, cell culture supernatants and urine. HSP70 is an important ATP-dependent cytosolic chaperone along with Hsp90. Hspa4 aids in the folding of several proteins and helps in protecting against cellular stresses such as rise in temperature. HSP70 maintains the function of the mitotic centrosome to coordinates bipolar mitotic spindle assembly. Hspa4 might compensate the loss of parkin function in mice by protecting the cells against proteolytic and mitochondrial stress. Therefore, it can serve as an important therapeutic agent for parkinson disease. Upregulation of HSPA4 is observed in parkin gene knockout mice. Hspa4 is known to be associated with tumor progression and offers resistance against many therapeutic agents. The gene is upregulated in many different types of cancer.
Immunogen
Recombinant Human HSP70
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jie Hong et al.
Biological trace element research, 159(1-3), 269-277 (2014-04-17)
The widespread application of lanthanoids (Lns) in manufacturing industries has raised occupational and environmental health concerns about the possible increased health risks to humans exposed to Lns in their working and living environments. Numerous studies have shown that exposures to
Bing Leng et al.
Neuroscience letters, 723, 134866-134866 (2020-02-29)
Cognitive decline is a common non-motor symptom of Parkinson disease (PD), and cellular prion protein (PrPC) has been suggested to play a role in this process. This study aimed to investigate the correlation between plasma exosomal prion protein and cognitive
Daniel Alcolea et al.
Neurology, 89(2), 178-188 (2017-06-09)
To analyze the clinical utility of 3 CSF biomarkers and their structural imaging correlates in a large cohort of patients with different dementia and parkinsonian syndromes within the spectrum of frontotemporal lobar degeneration (FTLD). We analyzed 3 CSF biomarkers (YKL-40
Induction of Hsp70 in tumor cells treated with inhibitors of the Hsp90 activity: A predictive marker and promising target for radiosensitization
Kudryavtsev VA, et al.
PLoS ONE, 12(3), e0173640-e0173640 (2017)
Berrak Guven et al.
Regulatory toxicology and pharmacology : RTP, 104, 128-132 (2019-03-18)
Radiation induced colitis is one the most common clinical issue for patients receiving radiotherapy. For this reason, we aimed to investigate the effect of antioxidant-effective flavonoids hesperidin and quercetin on the intestinal damage induced by radiation in this study. TNF-alpha
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service