Skip to Content
Merck
All Photos(1)

Key Documents

RAB0210

Sigma-Aldrich

Mouse Granzyme B ELISA Kit

for serum, plasma and cell culture supernatant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 12 pg/mL
standard curve range: 14.34-3500 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Gzmb(14939)

General description

The Mouse Granzyme B ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse Granzyme B in serum, plasma, cell culture supernatants and urine.
The Mouse Granzyme B enzyme-linked immunosorbent assay (ELISA) kit is an in vitro assay for the quantitative measurement of mouse granzyme B in serum, plasma, cell culture supernatants and urine. Granzyme B binds to mannose-6-phosphate/insulin-like growth factor II receptor. This protein is implicated in fast DNA fragmentation, a key step towards apoptosis in cells. Granzyme B is involved in the cleaving of pro-caspase-3, which leads to the activation of caspase-3. This protein also cleaves caspase-6, -7, -8, -9, and -10.

Immunogen

Recombinant Mouse Granzyme B

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brendan Bulik-Sullivan et al.
Nature biotechnology (2018-12-18)
Neoantigens, which are expressed on tumor cells, are one of the main targets of an effective antitumor T-cell response. Cancer immunotherapies to target neoantigens are of growing interest and are in early human trials, but methods to identify neoantigens either
Kosei Hasegawa et al.
Journal of immunotherapy (Hagerstown, Md. : 1997), 41(4), 201-207 (2018-02-13)
Cancer immunotherapy has now been established as a leading standard therapeutic option in a subset of patients with cancer. In this study, we conducted a phase I dose-escalation trial using a mixture of 5 peptides to vaccinate cervical cancer patients
Ciputra Adijaya Hartana et al.
PloS one, 13(7), e0200079-e0200079 (2018-07-03)
The immune system plays a significant role in urothelial bladder cancer (UBC) progression, with CD8+ T cells being capable to directly kill tumor cells using perforin and granzymes. However, tumors avoid immune recognition by escape mechanisms. In this study, we
Patrick R Hagner et al.
British journal of haematology, 179(3), 399-409 (2017-08-05)
Lenalidomide is an immunomodulatory agent that has demonstrated clinical benefit for patients with relapsed or refractory mantle cell lymphoma (MCL); however, despite this observed clinical activity, the mechanism of action (MOA) of lenalidomide has not been characterized in this setting.
Hannah Jonescheit et al.
Cells, 9(5) (2020-05-10)
Pancreatic ductal adenocarcinoma (PDAC) is a malignant gastrointestinal disease. The enzyme indoleamine-2,3-dioxgenase (IDO) is often overexpressed in PDAC and its downstream metabolite kynurenine has been reported to inhibit T cell activation and proliferation. Since γδ T cells are of high

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service