Skip to Content
Merck
All Photos(1)

Key Documents

R4533

Sigma-Aldrich

RNAzol® RT

For processing total and small RNA from human, animal, plant, bacterial,and viral samples

Synonym(s):

single step RNA isolation, single step RNA purification, total RNA isolation, total RNA purification

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.52

Quality Level

usage

1 mL sufficient for 107 cells
1 mL sufficient for 100 mg tissue (or)

General description

RNAzol RT is a quick and convenient reagent for use in the single-step isolation of total and small RNA from biological samples of human, animal, plant, yeast, bacterial, and viral origin. A convenient single-step liquid phase separation results in the isolation of RNA from DNA, protein, polysaccharides, and other molecules. RNAzol® RT can be used to isolate separate fractions of mRNA and micro RNA or to isolate total RNA, containing all classes of RNA in a single fraction.

This product, a mixture of guanidine thiocyanate and phenol in a monophase solution, effectively dissolves DNA, RNA, and protein on homogenization or lysis of tissue sample. Addition of water to the mixture allows for the precipitation of DNA, proteins, polysaccharides and other molecules, which can then be removed by centrifugation. RNA can then be isolated by alcohol precipitation, washing and solubilization. Chloroform-induced phase separation is not necessary. One mL of RNAzol® RT is sufficient to isolate RNA from up to 100 mg of tissue or 10e7 cells.

This is one of the most effective methods for isolating total and small RNA and can be completed at room temperature in less than 1 hour starting with fresh tissue or cells. The procedure is very effective for isolating RNA molecules of all types: large nuclear RNA, rRNA, mRNA, small RNA and micro RNA. The resulting RNA is intact with little or no contaminating DNA and protein.

Application

Isolated RNA can be used for Northern blots, RNase protection assay, microarrays, PCR, and other molecular biology applications.
RNAzol RT is a highly effective reagent for the single-step isolation of total and small RNA from human, animal, plant, bacterial and viral origin samples. The RNA isolation method based on this reagent is performed at room temperature, without the use of chloroform for phase separation, and allows for use in RT-PCR without requiring DNase treatment.

Features and Benefits

  • Isolates micro RNA and total RNA isolation of pure and undegraded RNA from biological samples
  • High yields and increased quality of isolated RNA
  • Completed at room temperature in less than an hour starting with fresh tissue or cells

Legal Information

RNAZol is a registered trademark of Molecular Research Center, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

Target Organs

Nervous system,Kidney,Liver,Skin

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Miccoli et al.
Marine environmental research, 127, 137-147 (2017-04-17)
Natural/synthetic Endocrine Disrupting Chemicals (EDCs) may display estrogenic activity and a lower potency than 17β-estradiol. Nonetheless, their concentrations and additive effects can affect the endocrine system and reproductive processes related to the Hypothalamic-Pituitary-Gonadal (HPG) axis. Because of their persistence in
Arinder K Arora et al.
Genes, 12(3) (2021-04-04)
Insect pest control by RNA interference (RNAi)-mediated gene expression knockdown can be undermined by many factors, including small sequence differences between double-stranded RNA (dsRNA) and the target gene. It can also be compromised by effects that are independent of the
Paulina Andryka-Dudek et al.
International journal of molecular sciences, 20(5) (2019-03-01)
Short-term (3 h) treatment of embryos isolated from dormant apple (Malus domestica Borkh.) seeds with NO donors stimulates their transition from dormancy to germination. Seed dormancy is maintained by ABA, while germination is controlled mainly by gibberellins (GAs) and jasmonic
Arturo Vargas et al.
Zebrafish, 15(4), 404-419 (2018-03-29)
A desirable goal of the aquaculture sector is to replace most of fish meal and fish oil with more sustainable, cost-effective, and environmental friendly ingredients ensuring fish health and welfare standards. Due to minimal environmental impact, compared with most conventional
V Shilova et al.
Cell stress & chaperones, 25(2), 305-315 (2020-02-11)
Here, we monitored the expression of three genes (hsp70, hsp22, and hsf1) involved in heat shock response in Drosophila melanogaster in males and females of different age. Also, we investigated age- and sex-dependent expression of three major genes participating in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service