Skip to Content
Merck
All Photos(1)

Key Documents

PZ0362

Sigma-Aldrich

CP-640186 hydrochloride

≥95% (HPLC)

Synonym(s):

(3R)-Anthracen-9-yl-[3-(morpholine-4-carbonyl)-[1,4′]bipiperidinyl-1′-yl]-methanone hydrochloride, CP 640186 hydrochloride, CP640186 hydrochloride, [(3R)-1′-(9-Anthracenylcarbonyl)[1,4′-bipiperidin]-3-yl]-4-morpholinyl-methanone hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H35N3O3 · HCl
CAS Number:
Molecular Weight:
522.08
MDL number:
UNSPSC Code:
41121800
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

O=C(C1=C2C=CC=CC2=CC3=CC=CC=C31)N(CC4)CCC4N5C[C@H](C(N6CCOCC6)=O)CCC5.[H]Cl

InChI

1S/C30H35N3O3.ClH/c34-29(32-16-18-36-19-17-32)24-8-5-13-33(21-24)25-11-14-31(15-12-25)30(35)28-26-9-3-1-6-22(26)20-23-7-2-4-10-27(23)28;/h1-4,6-7,9-10,20,24-25H,5,8,11-19,21H2;1H/t24-;/m1./s1

InChI key

DUBNXJIOBFRASV-GJFSDDNBSA-N

Application

CP-640186 hydrochloride has been used as an allosteric acetyl-CoA carboxylase (ACC) inhibitor (ACCi) for pharmacological inhibition of lipogenesis to determine if fatty acid synthesis is essential for brown adipose tissue (BAT) whitening. It has also been used as an inhibitor of mammalian acetyl-CoA carboxylase (mACC1/2) to study its effect on meropenem potency against the persisters from multiple pathogens, including B. thailandensis, P. aeruginosa, S.Typhimurium, and attenuated Y. pestis.

Biochem/physiol Actions

CP-640186 is a potent and orally active acetyl-CoA carboxylase 1/2 (ACC-alpha/beta, ACC1/2) inhibitor (IC50 ~50 nM) that targets the carboxyltransferase (CT) domain at the ACC dimer interface (via tight interactions with the putative biotin-binding site) in a reversible manner, uncompetitive with respect to ATP, and non-competitive with respect to bicarbonate, acetyl-CoA, and citrate. CP-610431 inhibits fatty acid (FA) synthesis, triglyceride (TG) synthesis, TG and apoB secretion (IC50 = 1.6, 1.8, 3.0, and 5.7 μM, respectively), but not cholesterol synthesis or apoC3 secretion in HepG2 cells (ACC1), as well as stimulates FA oxidation in C2C12 cells (ACC2) and in rat epitrochlearis muscle strips (EC50 = 57 nM and 1.3 μM, respectively). Oral administration is shown to inhibit FA synthesis in rats, CD1 mice, and ob/ob mice (ED50 = 13, 11, and 4 mg/kg, respectively) and stimulate rat whole body FA oxidation (ED50 ∼30 mg/kg) in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hailong Zhang et al.
Structure (London, England : 1993), 12(9), 1683-1691 (2004-09-03)
Acetyl-coenzyme A carboxylases (ACCs) are important targets for the development of therapeutic agents against obesity, diabetes, and other diseases. CP-640186 is a potent inhibitor of mammalian ACCs and can reduce body weight and improve insulin sensitivity in test animals. It
Deepa S Valsangkar et al.
Molecular reproduction and development, 82(9), 679-693 (2015-06-05)
In mouse oocytes, meiotic induction by pharmacological activation of PRKA (adenosine monophosphate-activated protein kinase; formerly known as AMPK) or by hormones depends on stimulation of fatty acid oxidation (FAO). PRKA stimulates FAO by phosphorylating and inactivating acetyl CoA carboxylase (ACAC;
P B Patil et al.
Archives of physiology and biochemistry, 113(1), 13-24 (2007-05-25)
There seems to be an association between increased concentrations of malonyl coenzyme A (malonyl CoA) in skeletal muscle and diabetes and/or insulin resistance. The purpose of the current study was to test the hypothesis that treatments designed to manipulate malonyl
Kevin P Madauss et al.
Acta crystallographica. Section D, Biological crystallography, 65(Pt 5), 449-461 (2009-04-25)
Inhibition of acetyl-CoA carboxylase (ACC) may prevent lipid-induced insulin resistance and type 2 diabetes, making the enzyme an attractive pharmaceutical target. Although the enzyme is highly conserved amongst animals, only the yeast enzyme structure is available for rational drug design.
Yasunori Nio et al.
Antiviral research, 132, 262-267 (2016-07-10)
Recently, direct antiviral agents against hepatitis C virus (HCV) infection have been developed as highly effective anti-HCV drugs. However, the appearance of resistant viruses against direct anti-viral agents is an unsolved problem. One of the strategies considered to suppress the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service