Skip to Content
Merck
All Photos(1)

Documents

PZ0195

Sigma-Aldrich

CYP3cide

≥98% (HPLC)

Synonym(s):

1-Methyl-3-[1-methyl-5-(4-methylphenyl)-1H-pyrazol-4-yl]-4-[(3S)-3-piperidin-1-ylpyrrolidin-1-yl]-1H-pyrazolo[3,4-d]pyrimidine, PF-04981517

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H32N8
CAS Number:
Molecular Weight:
456.59
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: >10 mg/mL (warmed)

storage temp.

room temp

SMILES string

Cc1ccc(cc1)-c2c(cnn2C)-c3nn(C)c4ncnc(N5CC[C@@H](C5)N6CCCCC6)c34

InChI

1S/C26H32N8/c1-18-7-9-19(10-8-18)24-21(15-29-31(24)2)23-22-25(32(3)30-23)27-17-28-26(22)34-14-11-20(16-34)33-12-5-4-6-13-33/h7-10,15,17,20H,4-6,11-14,16H2,1-3H3/t20-/m0/s1

InChI key

WDWIMDKOXZZYHH-FQEVSTJZSA-N

Biochem/physiol Actions

CYP3cide is a mechanism-based inhibitor of cytochrome P4503A4 that can be used to distinguish the contributions of CYP3A4 versus CYP3A5 on drug metabolism. The IC50 values for inhibition of Midazolam 1′-Hydroxylase activity of recombinant CYP3A4 and CYP3A5 by CYP3cide are 0.3 and 17 mM, respectively.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Legal Information

Sold for research purposes under agreement from Pfizer Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew Rowland et al.
British journal of clinical pharmacology, 85(1), 216-226 (2018-10-20)
Demonstrate the presence of cytochrome P450 (CYP) and UDP-glucuronosyltransferase (UGT) proteins and mRNAs in isolated human plasma exosomes and evaluate the capacity for exosome-derived biomarkers to characterize variability in CYP3A4 activity. The presence of CYP and UGT protein and mRNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service