Skip to Content
Merck
All Photos(1)

Key Documents

PRS4877

Sigma-Aldrich

Anti-IL-17 (ab1) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CTLA8, Anti-Interleukin 17A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 17 kDa

species reactivity

mouse, human

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL17A(3605)

General description

IL-17A (interleukin 17A) is a proinflammatory cytokine of Th17 (helper T cells). It is located on chromosome 6p12.

Immunogen

a 16 amino acid peptide from near the amino-terminus of human IL-17A.

Application

Anti-IL-17 has been used in western blotting and immunohistochemistry.

Biochem/physiol Actions

IL-17A (interleukin 17A) plays a major role in the pathogenesis of autoimmune diseases like multiple sclerosis and rheumatoid arthritis. It also plays a pathogenic role in allergic rhinitis (AR). It helps in the formation of several chemokines and cytokines in airway epithelial cells.

Linkage

The action of this antibody can be blocked using blocking peptide SBP4877.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

IL-17A and Th17 cells in lung inflammation: an update on the role of Th17 cell differentiation and IL-17R signaling in host defense against infection
Tsai HC, et al.
Clinical & Developmental Immunology (2013)
Nicola Tamassia et al.
European journal of clinical investigation, 48 Suppl 2, e12952-e12952 (2018-05-18)
Polymorphonuclear neutrophils are the most numerous leucocytes present in human blood, and function as crucial players in innate immune responses. Neutrophils are indispensable for the defence towards microbes, as they effectively counter them by releasing toxic enzymes, by synthetizing reactive
Nicola Tamassia et al.
Frontiers in immunology, 9, 795-795 (2018-05-03)
Neutrophils are known to perform a series of effector functions that are crucial for the innate and adaptive responses, including the synthesis and secretion of a variety of cytokines. In light of the controversial data in the literature, the main
Foxp3 inhibits RORgammat-mediated IL-17A mRNA transcription through direct interaction with RORgammat
Ichiyama K, et al.
The Journal of Biological Chemistry, 283(25), 17003-17008 (2008)
Association between polymorphisms in cytokine genes IL-17A and IL-17F and development of allergic rhinitis and comorbid asthma in Chinese subjects
Wang M, et al.
Human immunology, 73(6), 647-653 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service