Skip to Content
Merck
All Photos(3)

Key Documents

PRS3363

Sigma-Aldrich

Anti-ST2 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Interleukin 1 receptor-like 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Il1rl1(17082)

General description

ST2 (suppression of tumorigenicity) belongs to the IL-1 family of cytokines. It is also known as IL-33 (interleukin 33) and IL1RL1 (interleukin 1 receptor like 1). It is expressed in macrophages and various inflammatory cells like T helper 2 (Th2), mast cells, basophils and eosinophils. It is mapped to chromosome 2q12.

Immunogen

a synthetic peptide corresponding to 16 amino acids at the amino-terminus of mouse ST2. This peptide is common to all three known ST2 isoforms.

Application

Anti-ST2 has been used in immunohistochemistry, western blottingand immunofluorescence staining.

Biochem/physiol Actions

ST2 (suppression of tumorigenicity) participates in pathophysiology of several diseases like asthma, arthritis, obesity and atherosclerosis. It works as a mitogen in eosinophils, fibrocytes, endothelial cells. It helps in the migration of Th2 lymphocytes, fibrocytes and endothelial cells, and disturbs the incursion and metastasis of breast tumor.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3363.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mamdouh A K Sedhom et al.
Gut, 62(12), 1714-1723 (2012-11-23)
Inflammatory bowel diseases (IBD) have been intrinsically linked to a deregulated cytokine network, but novel therapeutic principles are urgently needed. Here we identify the interleukin (IL)-33 and its receptor ST2 as key negative regulators of wound healing and permeability in
ST2L Transmembrane Receptor Expression: An Immunochemical Study on Endarterectomy Samples
Marzullo A, et al.
PLoS ONE, 11(5) (2016)
The IL-33/ST2 pathway contributes to intestinal tumorigenesis in humans and mice
Mertz KD, et al.
Oncoimmunology, 5(1) (2015)
Wen-Ting Hu et al.
Molecular human reproduction, 20(4), 358-372 (2013-12-18)
Interleukin (IL)-33, a newly described member of the IL-1 family, has been reported to facilitate primary tumor progression and metastatic dissemination. However, its biological function on decidual stromal cells (DSCs) remains unclear. In this study, we tested the hypothesis whether
Mark Barbour et al.
Journal of neuroimmunology, 318, 87-96 (2018-03-13)
Experimental autoimmune encephalomyelitis (EAE) mice were administered with murine anti-CD52 antibody to investigate its therapeutic effect and whether the treatment modulates IL-33 and ST2 expression. EAE severity and central nervous system (CNS) inflammation were reduced following the treatment, which was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service