Skip to Content
Merck
All Photos(4)

Key Documents

PRS3335

Sigma-Aldrich

Anti-Bcl-2 (ab1) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-B-cell CLL/lymphoma 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BCL2(596)

Related Categories

General description

B-cell lymphoma 2 (BCL2), a proto-oncogene is made of three exons. It is widely expressed in immature B-cells and memory B cells. BCL2 is found at high levels in the thymus, spleen, and lymph nodes. BCL2 gene is mapped to human chromosome 18q21.33.

Immunogen

a peptide corresponding to 15 amino acids near the N-terminus of human Bcl-2.

Application

Anti-Bcl-2 (ab1) antibody produced in rabbit has been used in western blotting (1:100) (1:1000), immunohistochemistry (1:50), immunoprecipitation (IP) (1:1000)

Biochem/physiol Actions

B-cell lymphoma 2 (BCL2) plays a key role in maintaining the integrity of the mitochondrial membrane. It blocks the release of cytochrome C and its binding to apoptosis activating factor-1 (APAF1). BCL2 family proteins participate in modulating cell death. It also regulates apoptosis, necrosis, and autophagy. Upregulation of BCL2 is seen in several human cancers. Overexpression of BCL2 is observed in chronic lymphocytic leukemia (CLL), marginal cell lymphoma, and mantle cell lymphoma.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3335.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keqin Chen et al.
Cardiovascular therapeutics, 2022, 3889419-3889419 (2022-12-08)
Small heat shock protein-1 (HSPB1) is a small heat shock protein that participates in many cellular processes and alleviates stress-induced cell injury. Autophagy protects cells from many types of stress and plays a key role in preventing stress in arteriosclerosis
Janani Ravikrishnan et al.
Haematologica (2024-08-08)
Patients with chronic lymphocytic leukemia (CLL) respond well to initial treatment with the Bcell lymphoma 2 (BCL2) inhibitor venetoclax. Upon relapse, they often retain sensitivity to BCL2 targeting, but durability of response remains a concern. We hypothesize that targeting both
Ping He et al.
International journal of molecular medicine, 45(4), 1261-1269 (2020-03-04)
The authors' previous studies demonstrated that the major renal damage from hepatitis B virus infection is HBx‑induced apoptosis of renal tubular epithelial cells. Cordyceps sinensis is one of the most valuable of traditional Chinese medicines and is extensively used to treat
Yudi Han et al.
International journal of molecular medicine, 41(2), 800-808 (2017-12-06)
Autophagy may be a major mechanism by which osteoblasts (OBs) protect against the negative effects of chronic glucocorticoid (GC) usage. OBs are closely associated with the remodeling that occurs in GC‑induced osteoporosis (GIO). In osteocytes, in response to stress induced by GCs, several
Leilei Yu et al.
Experimental and therapeutic medicine, 12(4), 2275-2280 (2016-10-05)
Diabetes is a metabolic disease that results from impairment in insulin secretion. The present study aimed to investigate the potential role of NOM1 in the function of pancreatic islet β cells and insulin secretion. MIN6 cells isolated from mice were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service