Skip to Content
Merck
All Photos(2)

Key Documents

PLA0300

Sigma-Aldrich

Goat anti-CRSP1/TRAP220 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

ARC205, CRSP1, CRSP200, DRIP205, DRIP230, Mediator complex subunit 1, PBP, PPAR binding protein, PPAR-binding protein, PPARBP, PPARG binding protein, PPARGBP, RB18A, TR-interacting protein 2, TRAP220, TRIP-2, TRIP2, activator-recruited cofactor 205 kDa component, p53 regulatory protein RB18A, peroxisome proliferator-activated receptor-binding protein, thyroid hormone receptor-associated protein complex 220 kDa component, thyroid hormone receptor-associated protein complex component TRAP220, thyroid receptor interacting protein 2, thyroid receptor-interacting protein 2, vitamin D receptor-interacting protein 230 kD, vitamin D receptor-interacting protein complex component DRIP205

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human, mouse

concentration

1 mg/mL

technique(s)

immunohistochemistry: 1:500-1:2,000
immunoprecipitation (IP): 2- 10 μg/mg
western blot: 1:500- 1:2,500

accession no.

NP_004765.2

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

Immunogen

The epitope recognized by PLA0300 maps to a region between residue 1531 and 1581 of human Cofactor Required for Sp1 Transcriptional Activation 1/Thyroid Hormone Receptor-Associated Protein Complex Component using the numbering given in entry NP_004765.2 (GeneID 5469).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

CSP1/TRAP220 is a component of the Mediator complex, a coactivator involved in the regulated transcription of nearly all RNA polymerase II-dependent genes. Mediator functions as a bridge to convey information from gene-specific regulatory proteins to the basal RNA polymerase II transcription machinery. Mediator is recruited to promoters by direct interactions with regulatory proteins and serves as a scaffold for the assembly of a functional preinitiation complex with RNA polymerase II and the general transcription factors [taken from the Universal Protein Resource (UniProt) www.uniprot.org/uniprot/Q15648].

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service